Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Healthcare activities protect and save lives, but what about the waste and by-products that they generate? Biomedical waste management can be expensive, and managing the potential risks associated with the prevention of infection, sharps-related hazards, and the environmental impact of treatment and disposal can be a major challenge.
Sterilwave is an innovative on-site solution for Biomedical Waste Management recommended by the World Health Organisation (WHO).
Sterilwave 100 | Sterilwave 250 | Sterilwave 440 | |
![]() | ![]() | ![]() | |
Particularly suited to medical facilities generating only small amounts of waste. Ultra-compact and comes in a mobile version, for use in a container for example. | Allows medium-sized institutions to treat their biomedical waste, guaranteeing safety and traceability at greatly reduced operating costs. | The ideal treatment solution for institutions producing a large quantity of biomedical waste, with minimal operating costs. | |
Waste capacity | 200 L/h–up to 20 kg/h (with a density of 1 L = 0.1 kg) | 500 L/h–up to 50 kg/h (with a density of 1 L = 0.1 kg) | 880 L/h–up to 70 kg/h (with a density of 1 L = 0.1 kg) |
Vessel capacity | 100 L | 250 L | 440 L |
Average cycle time | 30 minutes | 30 minutes | 30 – 32 minutes |
Dimensions ( L x W x D ) | 1.7 x 1.2 x 1.1 m | 1.6 x 1.1 x 1.5 m | 2 .0 x 1.1 x 2.0 m |
Weight | 700 kg | 1,200 kg | 1,400 kg |
Recommended working area | 10 sq m Ceiling height 2 m | 10 sq m (full workflow area) Ceiling height 2.2 m | 10 sq m (full workflow area) Ceiling height 3 m (3.85 m with automatic loading option) |
Electricity | 400 V/ 3-Phase, 40 A | 400 V/ 3-Phase, 63 A | 400 V/ 3-Phase, 63 A |
Technology | Grinding by rotating blades and heating by microwaves | ||
Volume reduction | Up to 80% | ||
Weight reduction | Up to 25% |