Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Linking the microstructure to the mechanical behavior is critical to the design of high-performance components for industry. 3D imaging techniques, such as lab/synchrotron X-ray tomography, optical tomography (OCT, OST), and MRI, allow you to perform in situ experiments with high spatial (down to the micron scale) and temporal resolution (within seconds). These techniques are particularly suitable for capturing complex and very short-lived phenomena, such as localization induced by heterogeneities, thermal mismatch between constituents, micro-cracking, fatigue behavior, and phase transitions. Digital volume correlation (DVC) exploits the natural texture of materials and has recently emerged as a powerful contactless, bulk strain measurement technique in experimental mechanics, materials science, and biomechanics.
Thermo Scientific Amira and Avizo Software are all-in-one image analysis platforms that allow you to compute 3D full-field displacement and strain maps from volume images acquired during the deformation process of an object. The data can be visualized and analyzed in a sophisticated way to quantify deformation-induced microstructural changes, such as strain transfer in multiphase materials, pore growth/coalescence related to fracture, and crack opening displacement (COD).
The damage mechanisms of polyamide 6 (PA6) matrix reinforced with unidirectional glass fibers have been characterized using synchrotron radiation laminography on CT-like specimens.
Digital volume correlation (DVC) enables researchers to go one step further to quantify in more details the 3D interaction of a crack with the microstructure.
Data courtesy of L. Laiarinandrasana & T. Morgeneyer from Centre des Matériaux Mines ParisTech and L. Helfen from the European Synchrotron Radiation Facility; Karlsruhe Institute of Technology.
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.
Leverage your 4D in-situ experiments using Avizo Software for Digital Volume Correlation.
Pore coalescence related to fracture of a fiber-reinforced polymer composite.
Analysis of 10ppi Aluminum open cell foam under compression. The Digital Volume Correlation analysis is based on the first three loading steps, showing the initial strain map distribution in the specimen. Data courtesy of TWI Ltd.
Avizo Software for Digital Volume Correlation to understand the dessication process in Egyptian crocodile mummies.
Leverage your 4D in-situ experiments using Avizo Software for Digital Volume Correlation.
Pore coalescence related to fracture of a fiber-reinforced polymer composite.
Analysis of 10ppi Aluminum open cell foam under compression. The Digital Volume Correlation analysis is based on the first three loading steps, showing the initial strain map distribution in the specimen. Data courtesy of TWI Ltd.
Avizo Software for Digital Volume Correlation to understand the dessication process in Egyptian crocodile mummies.