Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Thermo Scientific HyPerforma Glass Bioreactors offer easy operation, rapid assembly, and are manufactured with the highest technology and design standards. These benchtop glass bioreactors are developed to ensure sterile processes and minimize lost batches. Using a computational fluid dynamics (CFD) simulator, the HyPerforma Glass Bioreactor impellers provide optimal stirred-tank mixing with minimum shear force, resulting in a higher average kLa and better cell growth.
Controllers that provide ease of transferability and scale-up throughout your process with flexibility and open-architecture.
A range of intuitive process sensors to help you monitor processes, reduce failures, and gain efficiencies.
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: