Miele Laboratory Glassware Washers
Make your lab life easier
Learn more about Miele glasswashers
Guaranteed hygienically clean glassware
Thermo Fisher Scientific has partnered with Miele Professional to offer a high-quality solution for laboratory glassware used in analytical experiments. This solution is designed to be gentle on materials and is endorsed by top manufacturers of laboratory glassware. With the addition of specific accessories tailored to different models and applications, Miele Professional laboratory washers enable efficient and easy reprocessing of various types of glassware in laboratories.
Features
Miele’s laboratory glassware washers feature:
  • Powerful circulation pump: Powerful circulation pump ensures optimum cleaning results
  • EcoDry: Allows the load to dry and cool down faster
  • Waterproof protection system as standard: Providing piece of mind and assurance against flooding
  • Built in steam condenser: Assuring protection from steam vapors and aerosols
  • Environmental benefits: Excellent sound and heat insulation and recyclable materials used ensuring sustainability with lower running costs
  • Built in water softener: Assuring optimum cleaning results whatever your water hardness
  • Redundant temperature monitoring: In line with ISO EN 15883 assuring accurate temperature monitoring
  • Acid rinse: PG8583, PG8593 and PG89583 CD have a dosing pump for an acid wash and neutralisation
  • Demineralised water connections: All machines can be connected to a demineralised water system
  • IQ/OQ: IQ/OQ can be performed by our fully training service department and a performance verification is available
Explore the Miele range
PG 8504 Laboratory Glasswasher
Laboratory glass washer designed to clean glassware in schools, industrial, environmental and research laboratories. Suitable as a freestanding or under counter unit, heat and noise emissions have been reduced to a minimum. The PG 8504 has been designed with convenience in mind, with in-door salt container, reservoir for neutralising agent or rinsing agent and powder dispenser for cleaning agent.
Key features:
  • 6 programmes + 2 free programmable positions
  • Demineralized water connection with feed pump
  • Maximum rinse temperature 70°C
  • Wash up to 64 narrow-necked glassware with injectors*
  • Option for external liquid dispensing (DOS G80 or DOS G80/1)
  • Multi-stage filtration system
  • Flexible range of baskets and inserts
  • Single phase 15 amp power (plug included)
  • Tested for 15,000 operating hours
  • Conversion option to high pressure demin water supply available at an extra cost
*Injector modules can be washed on a single level only
PG 8583 Laboratory Glasswasher
The PG 8583 is a universal thermo-disinfector as it has a large wash cabinet and short programme cycles. After an intensive main cleaning phase, the disinfection takes place at >90oC and a holding time of 5 minutes. The final rinsing is carried out with demineralised water without rinsing agents. Each cycle can wash up to 128 laboratory flasks or 98 pipettes together with other laboratory glassware.
Key features:
  • Programmable, 14 programmes
  • Demineralized water connection
  • Maximum rinse temperature 93°C
  • Capacity 128 flasks or 98 pipettes
  • 2 integrated dispenser pumps: 1x detergent, 1x neutralizing agent
  • Multi-stage filtration system
  • Spray arm and spray pressure monitoring
  • A0 value control
  • EcoDry; convenient drying support
  • 3 phase power, convertible to single phase (charges apply)
  • Tested for 15,000 operating hours
  • Optional demineralized water with feed pump (flow pressure min 60kpa - max 1000kpa)
  • PG 8583 [WW AD LD OIL] laboratory washer also available with AD water connection, liquid dispensing, EcoDry function, and oil seals. Ideal for customers washing a lot of products with oil residues.
PG 8593 Laboratory Glasswasher
The PG 8593 is a 60 cm wide washer disinfector with integrated hot-air drying. An integrated HEPA filter (Class H13) guarantees the purity of the air used for drying. Intricately shaped glassware and instruments can be cleaned, disinfected and dried effectively through each cycle. The PG 8593 can process 128 laboratory flasks or 98 pipettes together with other laboratory glassware.
Key features:
  • Programmable, 15 programmes
  • Demineralized water connection
  • Maximum rinse temperature 93°C
  • Capacity 128 flasks or 98 pipettes
  • 1 integrated dispenser pump for liquid cleaning agent
  • Multi-stage filtration system
  • Spray arm and spray pressure monitoring
  • A0 value control
  • DryPlus with H13 HEPA filter
  • 3 phase power, convertible to single phase (charges apply)
  • Tested for 15,000 operating hours
PG 8583CD Laboratory Glasswasher
The PG 8583 CD is a 90 cm wide washer disinfector with integrated hot-air drying. An integrated HEPA filter (Class H 14) guarantees the purity of the air used for drying. Intricately shaped glassware and instruments can be cleaned, disinfected and dried effectively through each cycle. The PG 8583 CD can process up to 128 laboratory flasks or 98 pipettes together with other laboratory glassware. It has a side cabinet for 5l supply canisters.
Key features:
  • Programmable, 14 programmes
  • Demineralized water connection
  • Maximum rinse temperature 93°C
  • Capacity 128 flasks or 98 pipettes
  • 2 integrated dispenser pumps. 1x detergent, 1x neutralizing agent
  • Multi-stage filtration system
  • Spray arm and spray pressure monitoring
  • Conductivity monitoring
  • A0 value control
  • DryPlus with H14 HEPA filter
  • Drawer for two 5L canisters
  • 3 phase power, convertible to single phase (charges apply)
  • Tested for 15,000 operating hours
  • Conversion option to high pressure demin water supply available at an extra cost
Accessories
Miele Professional Laboratory Glasswashers are offered in combination with accessories specific to models and applications to facilitate the fast and simple reprocessing of all types of glassware. 
  • Baskets and load carriers 
  • EasyLoad inserts and other components 
  • Injector nozzles 
  • Conversion kits 
  • Transfer trolley 
  • Systems for reprocessing with fully demineralised water 
  • Dispensing components 
  • Plinths
Resources
Videos
Miele glasswashers benefits