Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003258.4 | NP_003249.3 | 7 | 1452 | ||
XM_017024992.1 | XP_016880481.1 | 6 | 1555 | |||
GenBank mRNA | BC006484.1 | - | 7 | 1282 | 2905608 | |
BC007872.2 | - | 7 | 1269 | 4303880 | ||
BC007986.2 | - | 8 | 898 | 2966331 | ||
K02581.1 | - | 7 | 1299 |