Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000544.3 | NP_000535.3 | 5 | 1032 | ||
NM_001290043.1 | NP_001276972.1 | 5 | 1085 | |||
NM_018833.2 | NP_061313.2 | 5 | 1032 | |||
GenBank mRNA | AB073779.1 | - | 4 | 910 | ||
AB208953.1 | - | 5 | 979 | |||
AF078671.1 | - | 5 | 989 | |||
AF105151.1 | - | 5 | 989 | |||
AK222823.1 | - | 4 | 941 | |||
AK223300.1 | - | 5 | 992 | |||
AK225657.1 | - | 5 | 1013 | |||
AK225693.1 | - | 5 | 992 | |||
AK292963.1 | - | 5 | 1015 | |||
BC002751.2 | - | 4 | 918 | |||
BT009906.1 | - | 4 | 910 | |||
FM246456.1 | - | 4 | 910 | |||
M74447.1 | - | 5 | 1006 | |||
M84748.1 | - | 5 | 938 | |||
U07844.1 | - | 4 | 720 | |||
Z22935.1 | - | 5 | 938 | |||
Z22936.1 | - | 5 | 986 |