Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000544.3 | NP_000535.3 | 6 | 1180 | ||
NM_001290043.1 | NP_001276972.1 | 6 | 1233 | |||
NM_018833.2 | NP_061313.2 | 6 | 1180 | |||
GenBank mRNA | AB073779.1 | - | 5 | 1058 | ||
AB208953.1 | - | 6 | 1127 | |||
AF078671.1 | - | 6 | 1137 | |||
AF105151.1 | - | 6 | 1137 | |||
AF176984.1 | - | 1 | 113 | |||
AK222823.1 | - | 5 | 1089 | |||
AK223300.1 | - | 6 | 1140 | |||
AK225657.1 | - | 6 | 1161 | |||
AK225693.1 | - | 6 | 1140 | |||
AK292963.1 | - | 6 | 1163 | |||
BC002751.2 | - | 5 | 1066 | |||
BT009906.1 | - | 5 | 1058 | |||
FM246456.1 | - | 5 | 1058 | |||
M74447.1 | - | 6 | 1154 | |||
M84748.1 | - | 6 | 1086 | |||
U07844.1 | - | 5 | 868 | |||
Z22935.1 | - | 6 | 1086 | |||
Z22936.1 | - | 6 | 1134 |