Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003258.4 | NP_003249.3 | 2 | 294 | ||
XM_017024992.1 | XP_016880481.1 | 2 | 298 | |||
GenBank mRNA | AB451268.1 | - | 2 | 84 | ||
AB451395.1 | - | 2 | 84 | |||
AK314950.1 | - | 2 | 137 | |||
BC006484.1 | - | 2 | 124 | 2905608 | ||
BC007872.2 | - | 2 | 111 | 4303880 | ||
BC007986.2 | - | 2 | 120 | 2966331 | ||
BI754079.1 | - | 2 | 324 | |||
BT006941.1 | - | 2 | 84 | |||
K02581.1 | - | 2 | 141 |