Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244938.1 | NP_001231867.1 | 4 | 452 | ||
NM_003329.3 | NP_003320.2 | 5 | 512 | |||
GenBank mRNA | AF065241.1 | - | 4 | 244 | ||
AF085844.1 | - | 5 | 319 | |||
AF276919.1 | - | 4 | 307 | |||
AF313911.1 | - | 5 | 317 | |||
AK289508.1 | - | 5 | 433 | |||
AW160756.1 | - | 5 | 512 | |||
AY004872.1 | - | 5 | 317 | |||
BC003377.1 | - | 5 | 364 | 2901084 | ||
BC054866.1 | - | 5 | 383 | 6463280 | ||
BX647247.1 | - | 4 | 353 | |||
J04026.1 | - | 5 | 370 | |||
JQ313905.1 | - | 5 | 307 | |||
X77584.1 | - | 5 | 387 |