Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGMAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024758.4 | NP_079034.3 | 6 | 1093 | ||
GenBank mRNA | AK130034.1 | - | 6 | 889 | ||
AK225366.1 | - | 6 | 1006 | |||
AY057097.1 | - | 6 | 993 | |||
BC005090.1 | - | 6 | 1093 | 3837251 |