Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000143.3 | NP_000134.2 | 10 | 1551 | ||
XM_011544132.2 | XP_011542434.1 | 10 | 1991 | |||
GenBank mRNA | AK312415.1 | - | 10 | 1527 | ||
BC003108.1 | - | 10 | 1513 | 3502012 | ||
BC017444.1 | - | 10 | 1503 | 3606118 | ||
BT009839.1 | - | 10 | 1488 | |||
KU178071.1 | - | 10 | 1488 | |||
KU178072.1 | - | 6 | 662 | |||
KU178074.1 | - | 4 | 453 | |||
KU178075.1 | - | 4 | 423 | |||
KU178076.1 | - | 5 | 369 | |||
M15502.1 | - | 9 | 1362 | |||
U48857.1 | - | 10 | 1518 | |||
U59309.1 | - | 10 | 1521 |