Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000802.3 | NP_000793.1 | 4, 5 | 518 | ||
NM_016724.2 | NP_057936.1 | 5, 6 | 769 | |||
NM_016725.2 | NP_057937.1 | 4, 5 | 703 | |||
NM_016729.2 | NP_057941.1 | 3, 4 | 538 | |||
GenBank mRNA | AF000381.2 | - | 2, 3 | 186 | ||
AK223527.1 | - | 4, 5 | 557 | |||
BC002947.2 | - | 4, 5 | 650 | 3956659 | ||
BT007158.1 | - | 3, 4 | 493 | |||
CR542019.1 | - | 3, 4 | 493 | |||
J05013.1 | - | 4, 5 | 704 | |||
KU257686.1 | - | 3, 4 | 418 | |||
M25317.1 | - | 3, 4 | 433 | |||
M28099.1 | - | 5, 6 | 589 | |||
X62753.1 | - | 4, 5 | 923 |