Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000802.3 | NP_000793.1 | 2 | 122 | ||
NM_016724.2 | NP_057936.1 | 3 | 373 | |||
NM_016725.2 | NP_057937.1 | 2 | 307 | |||
NM_016729.2 | NP_057941.1 | 1 | 142 | |||
GenBank mRNA | AK223527.1 | - | 2 | 161 | ||
BC002947.2 | - | 2 | 254 | 3956659 | ||
BT007158.1 | - | 1 | 97 | |||
CR542019.1 | - | 1 | 97 | |||
J05013.1 | - | 2 | 308 | |||
KU257686.1 | - | 1 | 22 | |||
M25317.1 | - | 1 | 37 | |||
M28099.1 | - | 3 | 193 | |||
M35069.1 | - | 1 | 28 | |||
U78793.1 | - | 1 | 142 | |||
U78794.1 | - | 1 | 123 | |||
X62753.1 | - | 2 | 527 |