Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000802.3 | NP_000793.1 | 3 | 261 | ||
NM_016724.2 | NP_057936.1 | 4 | 512 | |||
NM_016725.2 | NP_057937.1 | 3 | 446 | |||
NM_016729.2 | NP_057941.1 | 2 | 281 | |||
GenBank mRNA | AK223527.1 | - | 3 | 300 | ||
BC002947.2 | - | 3 | 393 | 3956659 | ||
BT007158.1 | - | 2 | 236 | |||
CR542019.1 | - | 2 | 236 | |||
J05013.1 | - | 3 | 447 | |||
KU257686.1 | - | 2 | 161 | |||
M28099.1 | - | 4 | 332 | |||
X62753.1 | - | 3 | 666 |