Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002031.2 | NP_002022.1 | 1 | 737 | ||
XM_005266880.4 | XP_005266937.1 | 2 | 809 | |||
XM_005266881.2 | XP_005266938.1 | 2 | 726 | |||
XM_005266882.4 | XP_005266939.1 | 3 | 1038 | |||
XM_011535653.2 | XP_011533955.1 | 3 | 1041 | |||
XM_011535654.2 | XP_011533956.1 | 2 | 823 | |||
XM_011535655.2 | XP_011533957.1 | 2 | 820 | |||
XM_017010645.1 | XP_016866134.1 | 2 | 927 | |||
GenBank mRNA | AK313314.1 | - | 1 | 817 | ||
BC012916.1 | - | 1 | 582 | 4515877 | ||
U00803.1 | - | 1 | 737 | |||
U22322.1 | - | 1 | 699 |