Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003255.4 | NP_003246.1 | 5 | 2341 | ||
GenBank mRNA | AL110197.1 | - | 1 | 498 | ||
BC040445.1 | - | 4 | 2075 | |||
BC071586.1 | - | 5 | 2335 | 30337524 |