Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002333.3 | NP_002324.2 | 3, 4 | 356 | ||
XM_005258945.1 | XP_005259002.1 | 3, 4 | 273 | |||
GenBank mRNA | AB009462.1 | - | 3, 4 | 489 | ||
AK129963.1 | - | 2, 3 | 129 | |||
AK295505.1 | - | 2, 3 | 184 | |||
AK296536.1 | - | 3, 4 | 237 | |||
AK316314.1 | - | 2, 3 | 183 | |||
BC007408.2 | - | 3, 4 | 285 | 3009711 |