Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002355.3 | NP_002346.1 | 5 | 848 | ||
XM_005253376.2 | XP_005253433.1 | 5 | 710 | |||
GenBank mRNA | AK222792.1 | - | 5 | 729 | ||
AK291143.1 | - | 5 | 694 | |||
AK312689.1 | - | 5 | 733 | |||
BC017111.1 | - | 3 | 248 | |||
BC024206.1 | - | 5 | 740 | 3530887 | ||
CR457412.1 | - | 4 | 573 | |||
M16985.1 | - | 5 | 718 |