Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207024.1 | NP_001193953.1 | 2 | 378 | ||
NM_002355.3 | NP_002346.1 | 2 | 378 | |||
XM_005253376.2 | XP_005253433.1 | 2 | 240 | |||
GenBank mRNA | AK222792.1 | - | 2 | 259 | ||
AK291143.1 | - | 2 | 224 | |||
AK312689.1 | - | 2 | 263 | |||
BC024206.1 | - | 2 | 270 | 3530887 | ||
BU166950.1 | - | 2 | 273 | |||
CR457412.1 | - | 1 | 103 | |||
DC341018.1 | - | 2 | 378 | |||
M16985.1 | - | 2 | 248 |