Search Thermo Fisher Scientific
We have recently updated our acclaimed Applied Biosystems 3500 and 3500 xL Dx Genetic analyzers to comply with the IVDR.
Clients previously frustrated that these powerful devices were not available in the European Union can breathe easy knowing that their new IVDR-compliant Applied Biosystems Genetic Analyzer is only an order away.
The Applied Biosystems 3500 Dx series genetic analyzers with 3500 Dx Series Data Collection Software 3 IVD v3.0.1 deliver flexibility by offering dual-mode capability within the same system to help meet the needs of today and tomorrow.
The secure Diagnostic Mode supports run modules that will perform both Sanger sequencing and fragment analysis.
To accommodate various demands on throughput, we offer two models*: the 8-capillary 3500 system for low throughput and 24-capillary 3500xL system for medium throughput, giving you even more flexibility to meet your lab's throughput needs.
It's accurate
The 3500 Dx Series Genetic Analyzers, using Sanger sequencing, are designed to deliver the accuracy you demand.
It's simple. It works the way you do.
Designed with you in mind, the 3500 Dx Genetic Analyzer gives you automated operation, easy-to-install consumables, electronic tracking of instrument performance and maintenance tasks, and integrated analysis software—so you can focus on the outcome.
It's flexible
The system comes in an 8-capillary version (3500 Dx) and a 24-capillary version (3500xL Dx) to align with your throughput needs. You can run the instrument in either IVD mode or RUO mode to develop other applications.
It's backed by award-winning support
Keeping your lab operational is critical. Our technical and field service support staff respond quickly to your calls, so that you can continue to respond to those who depend on you for results.
Putting you in the driver's seat
3500 Dx Data Collection Software offers user-friendly navigation and an intuitive dashboard design, including:
Snap in and run
Capillary array
Cathode buffer container
Anode buffer container
Performance optimized polymer
Operates on standard power
The 3500 Dx Series is easy to install and operate:
Feature | Benefit |
---|---|
Cleared for sequencing and fragment analysis Approved for sequencing | An instrument built in an ISO-certified facility |
RFID | Instrument automatically checks for the use of correct product and expiration dates |
IVD reagents/accessories | Tested and built to operate optimally with the 3500 Dx platform |
SAE | Security, audit, and electronic signature capability to help meet compliance to governing regulations |
8-capillary or 24-capillary models | Select the appropriate instrument to meet your laboratory throughput needs |
Flexibility | Both IVD and RUO modes to allow you to adhere to your lab's development requirements—sequencing or fragment analysis applications |
IVDR Compliance | 3500 Dx series genetic analyzers are compliant with the In Vitro Diagnostic Regulation (IVDR). |
There’s no time for downtime in your lab. With more than 1,000 trained professionals ready to assist you when you need it, our service solutions for Applied Biosystems instruments and applications help keep your lab up and running.
You’re covered
Every new instrument purchase comes with a one-year warranty. Longer-term service plans are also available that include guaranteed response times* and priority phone access to our trained and certified technical and instrument support specialists. When purchased at the same time as your instrument, these services plans can help maximize system uptime, reduce overall repair costs, get fast repair turnaround time, extend the life of your instrument, and help keep it running at peak performance.
Beyond repair to proactive care
The AB Assurance Plan is our premium repair plan, designed to maximize instrument performance and help ensure availability of critical systems with preventive maintenance, proactive instrument monitoring, remote diagnostic capabilities, and—should one of your instruments require repair—a fast response.
Qualification services
Whether you need a qualification performed with a new installation or after a qualifying service event, our qualification services provide documentation to verify that your instrument is performing according to manufacturer’s specifications.
Your instruments are now easier to manage
All your instrument use and care records are now in one place—the no-cost, no-hassle Instrument Management tool. Get instant access to the complete service history of your instruments, check instrument availability, schedule instrument time from anywhere in the world, and more.
Education services help enable your success
It can be difficult to prepare yourself for what’s next while you’re focused on the work you have now. Our professional, interactive training courses make it easier. We offer a combination of virtual and in-person classroom instruction, and hands-on learning in your lab to match your schedule, budget, and learning preferences. Whichever course style you choose, you’ll learn from one of our 300 highly skilled application scientists who are available to lead sessions online, at your location, or at one of our 12 training centers located worldwide.
*Guaranteed response times are dependent on type of service plan and geographic region.
AB Assurance with qualification services* | |
---|---|
On-site response time for repairs (business days) | Guaranteed 2 business days |
Remote instrument monitoring and diagnostics | Guaranteed 2 business days |
Priority phone and email access to instrument support | Guaranteed 2 business days |
Application technical support | Guaranteed 2 business days |
Instrument operating software updates | Guaranteed 2 business days |
Instrument qualification services | Guaranteed 2 business days |
Planned maintenance | Guaranteed 2 business days |
Parts, labor, and travel | Guaranteed 2 business days |
*Terms and conditions apply. |
It's accurate
The 3500 Dx Series Genetic Analyzers, using Sanger sequencing, are designed to deliver the accuracy you demand.
It's simple. It works the way you do.
Designed with you in mind, the 3500 Dx Genetic Analyzer gives you automated operation, easy-to-install consumables, electronic tracking of instrument performance and maintenance tasks, and integrated analysis software—so you can focus on the outcome.
It's flexible
The system comes in an 8-capillary version (3500 Dx) and a 24-capillary version (3500xL Dx) to align with your throughput needs. You can run the instrument in either IVD mode or RUO mode to develop other applications.
It's backed by award-winning support
Keeping your lab operational is critical. Our technical and field service support staff respond quickly to your calls, so that you can continue to respond to those who depend on you for results.
Putting you in the driver's seat
3500 Dx Data Collection Software offers user-friendly navigation and an intuitive dashboard design, including:
Snap in and run
Capillary array
Cathode buffer container
Anode buffer container
Performance optimized polymer
Operates on standard power
The 3500 Dx Series is easy to install and operate:
Feature | Benefit |
---|---|
Cleared for sequencing and fragment analysis Approved for sequencing | An instrument built in an ISO-certified facility |
RFID | Instrument automatically checks for the use of correct product and expiration dates |
IVD reagents/accessories | Tested and built to operate optimally with the 3500 Dx platform |
SAE | Security, audit, and electronic signature capability to help meet compliance to governing regulations |
8-capillary or 24-capillary models | Select the appropriate instrument to meet your laboratory throughput needs |
Flexibility | Both IVD and RUO modes to allow you to adhere to your lab's development requirements—sequencing or fragment analysis applications |
IVDR Compliance | 3500 Dx series genetic analyzers are compliant with the In Vitro Diagnostic Regulation (IVDR). |
There’s no time for downtime in your lab. With more than 1,000 trained professionals ready to assist you when you need it, our service solutions for Applied Biosystems instruments and applications help keep your lab up and running.
You’re covered
Every new instrument purchase comes with a one-year warranty. Longer-term service plans are also available that include guaranteed response times* and priority phone access to our trained and certified technical and instrument support specialists. When purchased at the same time as your instrument, these services plans can help maximize system uptime, reduce overall repair costs, get fast repair turnaround time, extend the life of your instrument, and help keep it running at peak performance.
Beyond repair to proactive care
The AB Assurance Plan is our premium repair plan, designed to maximize instrument performance and help ensure availability of critical systems with preventive maintenance, proactive instrument monitoring, remote diagnostic capabilities, and—should one of your instruments require repair—a fast response.
Qualification services
Whether you need a qualification performed with a new installation or after a qualifying service event, our qualification services provide documentation to verify that your instrument is performing according to manufacturer’s specifications.
Your instruments are now easier to manage
All your instrument use and care records are now in one place—the no-cost, no-hassle Instrument Management tool. Get instant access to the complete service history of your instruments, check instrument availability, schedule instrument time from anywhere in the world, and more.
Education services help enable your success
It can be difficult to prepare yourself for what’s next while you’re focused on the work you have now. Our professional, interactive training courses make it easier. We offer a combination of virtual and in-person classroom instruction, and hands-on learning in your lab to match your schedule, budget, and learning preferences. Whichever course style you choose, you’ll learn from one of our 300 highly skilled application scientists who are available to lead sessions online, at your location, or at one of our 12 training centers located worldwide.
*Guaranteed response times are dependent on type of service plan and geographic region.
AB Assurance with qualification services* | |
---|---|
On-site response time for repairs (business days) | Guaranteed 2 business days |
Remote instrument monitoring and diagnostics | Guaranteed 2 business days |
Priority phone and email access to instrument support | Guaranteed 2 business days |
Application technical support | Guaranteed 2 business days |
Instrument operating software updates | Guaranteed 2 business days |
Instrument qualification services | Guaranteed 2 business days |
Planned maintenance | Guaranteed 2 business days |
Parts, labor, and travel | Guaranteed 2 business days |
*Terms and conditions apply. |
Catalog # | Name | |
A27772 | Applied Biosystems 3500 Dx Genetic Analyzer (8-capillary) | Request for quote › |
A27856 | Applied Biosystems 3500xL Dx Genetic Analyzer (24-capillary) | Request for quote › |
*Instrument not available for sale in Morocco, Vietnam, Martinique, and Sri Lanka.
CE-IVD
For In Vitro Diagnostic Use.