Factors affecting pipette accuracy
Temperature
Temperature
Temperature has many effects on pipetting accuracy. The factor that has the greatest effect is the temperature difference between the delivery device and the liquid. The air gap (dead air volume) between the liquid surface and the piston experiences thermal expansion effects unique to the case. This either reduces or increases the liquid amount aspirated into the tip, along with other effects.
Density
Density
The density (mass/volume ratio) affects the liquid volume that is aspirated into the tip. A smaller dose of liquid with higher density than water is aspirated compared to a similar operation with water. With lower density liquids the effect is the opposite. This is caused by the flexible dead air volume along with earth’s gravity. The density of liquids also varies according to temperature. Typically the density of liquids also varies according to temperature. Typically the density for water is 0.998 kg/dm3, for ethanol 0.79 kg/dm3, and for sulfuric acid (95–98% h2SO4) 1.84 kg/dm3 (the values apply at the temperature of 20 °C/68 °F).
Altitude
Altitude
Geographic altitude affects accuracy through air pressure. Air pressure decreases in higher altitudes, and the conversion factor Z decreases as well. Also, with some liquids, the boiling point decreases quite close to room temperature, which will increase the evaporation loss dramatically.
Navigate this guide:
Liquid handling
Proper pipetting techniques
Ergonomics
Maintenance & service
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.