Product References
CARD11 gain-of-function mutation drives cell-autonomous accumulation of PD-1+ ICOShigh activated T cells, T-follicular, T-regulatory and T-follicular regulatory cells.
Frontiers in immunology
Masle-Farquhar E,Jeelall Y,White J,Bier J,Deenick EK,Brink R,Horikawa K,Goodnow CC
25-9985-82 was used in Flow cytometry/Cell sorting to reveal CARD11 as an important, cell-autonomous positive regulator of TFH, TREG and TFR cells.
Mon Mar 27 00:00:00 EDT 2023
Non-synergy of PD-1 blockade with T-cell therapy in solid tumors.
Journal for immunotherapy of cancer
Davies JS,Karimipour F,Zhang L,Nagarsheth N,Norberg S,Serna C,Strauss J,Chiou S,Gulley JL,Hinrichs CS
25-9985-82 was used in Flow cytometry/Cell sorting to support the concept that PD-1 blockade acts primarily through endogenous rather than transferred T cells to mediate a non-synergistic antitumor effect in solid tumor cell therapy.
Fri Jul 01 00:00:00 EDT 2022
Loss of Optineurin Drives Cancer Immune Evasion via Palmitoylation-Dependent IFNGR1 Lysosomal Sorting and Degradation.
Cancer discovery
Du W,Hua F,Li X,Zhang J,Li S,Wang W,Zhou J,Wang W,Liao P,Yan Y,Li G,Wei S,Grove S,Vatan L,Zgodziński W,Majewski M,Wallner G,Chen H,Kryczek I,Fang JY,Zou W
25-9985-82 was used in Flow Cytometry to show that optineurin is a shared node between the IFN and MHC pathways and predicts colorectal cancer patient outcome.
Thu Jul 01 00:00:00 EDT 2021
Recombinant programmed cell death 1 inhibits psoriatic inflammation in imiquimod‑treated mice.
International journal of molecular medicine
Peng S,Cao M,Sun X,Zhou Y,Chen CY,Ma T,Li H,Li B,Zhu B,Li X
25-9985-82 was used in Flow Cytometry to demonstrate that inhibition of PD‚1 function with antibodies promoted psoriasis development.
Sat Aug 01 00:00:00 EDT 2020
Targeting ANXA1 abrogates Treg-mediated immune suppression in triple-negative breast cancer.
Journal for immunotherapy of cancer
Bai F,Zhang P,Fu Y,Chen H,Zhang M,Huang Q,Li D,Li B,Wu K
25-9985 was used in Flow cytometry/Cell sorting to show that ANXA1 expression is associated with lower survival and a higher risk of breast malignancy.
Wed Apr 01 00:00:00 EDT 2020
Lymph Node Stromal Cells Generate Antigen-Specific Regulatory T Cells and Control Autoreactive T and B Cell Responses.
Cell reports
Nadafi R,Gago de Graça C,Keuning ED,Koning JJ,de Kivit S,Konijn T,Henri S,Borst J,Reijmers RM,van Baarsen LGM,Mebius RE
25-9985-82 was used in Flow Cytometry to uncover the ability of LNSCs to suppress the early activation of autoreactive immune cells and maintain peripheral tolerance.
Tue Mar 24 00:00:00 EDT 2020
The therapeutic significance of the novel photodynamic material TPE-IQ-2O in tumors.
Aging
Wang L,Liu Y,Liu H,Tian H,Wang Y,Zhang G,Lei Y,Xue L,Zheng B,Fan T,Zheng Y,Tan F,Xue Q,Gao S,Li C,He J
25-9985 was used in Flow cytometry/Cell sorting to explore the safety and effectiveness of a new mitochondria-targeting photodynamic material, TPE-IQ-2O, in combination therapies (combined with surgery or immunotherapy).
Sat Dec 19 00:00:00 EST 2020
A Thpok-Directed Transcriptional Circuitry Promotes Bcl6 and Maf Expression to Orchestrate T Follicular Helper Differentiation.
Immunity
Vacchio MS,Ciucci T,Gao Y,Watanabe M,Balmaceno-Criss M,McGinty MT,Huang A,Xiao Q,McConkey C,Zhao Y,Shetty J,Tran B,Pepper M,Vahedi G,Jenkins MK,McGavern DB,Bosselut R
25-9985-82 was used in Flow Cytometry to identify a transcriptional program that links the CD4+ lineage with Tfh differentiation, a limiting factor for efficient B cell responses.
Tue Sep 17 00:00:00 EDT 2019
The chemokine receptor CCR7 is a promising target for rheumatoid arthritis therapy.
Cellular & molecular immunology
Moschovakis GL,Bubke A,Friedrichsen M,Ristenpart J,Back JW,Falk CS,Kremmer E,Förster R
25-9985-82 was used in Flow Cytometry to demonstrate that CCR7 signaling is essential for the induction of CIA and identify CCR7 as a potential therapeutic target in RA.
Tue Oct 01 00:00:00 EDT 2019
PD-L1 in tumor microenvironment mediates resistance to oncolytic immunotherapy.
The Journal of clinical investigation
Zamarin D,Ricca JM,Sadekova S,Oseledchyk A,Yu Y,Blumenschein WM,Wong J,Gigoux M,Merghoub T,Wolchok JD
25-9985 was used in Flow cytometry/Cell sorting to use human tumour histocultures and syngeneic tumour models treated with Newcastle disease virus to identify immune targets upregulated by OV treatment.
Mon Apr 02 00:00:00 EDT 2018
Drp1 Controls Effective T Cell Immune-Surveillance by Regulating T Cell Migration, Proliferation, and cMyc-Dependent Metabolic Reprogramming.
Cell reports
Simula L,Pacella I,Colamatteo A,Procaccini C,Cancila V,Bordi M,Tregnago C,Corrado M,Pigazzi M,Barnaba V,Tripodo C,Matarese G,Piconese S,Campello S
25-9985 was used in Flow cytometry/Cell sorting to describe the crucial role of the mitochondrial protein Drp1 in T cell development and regulation of lymphocyte immune-surveillance.
Tue Dec 11 00:00:00 EST 2018
TIGIT and PD-1 dual checkpoint blockade enhances antitumor immunity and survival in GBM.
Oncoimmunology
Hung AL,Maxwell R,Theodros D,Belcaid Z,Mathios D,Luksik AS,Kim E,Wu A,Xia Y,Garzon-Muvdi T,Jackson C,Ye X,Tyler B,Selby M,Korman A,Barnhart B,Park SM,Youn JI,Chowdhury T,Park CK,Brem H,Pardoll DM,Lim M
25-9985 was used in Flow cytometry/Cell sorting to determine the effect of anti-PD-1 and anti-TIGIT combination therapy on survival in a murine glioblastoma model, and to elucidate the underlying immune mechanisms.
Wed Nov 03 00:00:00 EDT 2021
Lysis-independent potentiation of immune checkpoint blockade by oncolytic virus.
Oncotarget
Oseledchyk A,Ricca JM,Gigoux M,Ko B,Redelman-Sidi G,Walther T,Liu C,Iyer G,Merghoub T,Wolchok JD,Zamarin D
25-9985 was used in Flow cytometry/Cell sorting to study the immunogenic potential of an oncolytic virus in bladder cancer where existing immune checkpoint blockade has shown suboptimal response rates.
Tue Jun 19 00:00:00 EDT 2018
Oncolytic Adenovirus and Tumor-Targeting Immune Modulatory Therapy Improve Autologous Cancer Vaccination.
Cancer research
Jiang H,Rivera-Molina Y,Gomez-Manzano C,Clise-Dwyer K,Bover L,Vence LM,Yuan Y,Lang FF,Toniatti C,Hossain MB,Fueyo J
25-9985 was used in Flow cytometry/Cell sorting to demonstrate that combining an oncolytic virus with tumor-targeting immune checkpoint modulators elicits potent in situ autologous cancer vaccination, resulting in an efficacious, tumor-specific, and long-lasting therapeutic effect.
Sat Jul 15 00:00:00 EDT 2017
Loss of MAPK-activated protein kinase 2 enables potent dendritic cell-driven anti-tumour T cell response.
Scientific reports
Soukup K,Halfmann A,Dillinger B,Poyer F,Martin K,Blauensteiner B,Kauer M,Kuttke M,Schabbauer G,Dohnal AM
25-9985 was used in Flow cytometry/Cell sorting to suggest that MAPK-activated protein kinase 2 deletion leads to an expansion of stimulatory CD103+ dendritic cells, mounting a potent CD8+ T cell anti-tumour response in mice.
Mon Sep 18 00:00:00 EDT 2017
Assessing Immune-Related Adverse Events of Efficacious Combination Immunotherapies in Preclinical Models of Cancer.
Cancer research
Liu J,Blake SJ,Harjunpää H,Fairfax KA,Yong MC,Allen S,Kohrt HE,Takeda K,Smyth MJ,Teng MW
25-9985 was used in Flow cytometry/Cell sorting to develop a model that may be used to preclinically assess the therapeutic window of novel immunotherapy combinations in different tumor types.
Thu Sep 15 00:00:00 EDT 2016
Rapid linkage of innate immunological signals to adaptive immunity by the brain-fat axis.
Nature immunology
Kim MS,Yan J,Wu W,Zhang G,Zhang Y,Cai D
25-9985 was used in Flow cytometry/Cell sorting to investigate the role of the brain in innate immunological signals, showing rapid linkage of innate signals and adaptive immunity by the brain-fat axis.
Fri May 01 00:00:00 EDT 2015
Antigen presentation kinetics control T cell/dendritic cell interactions and follicular helper T cell generation in vivo.
eLife
Benson RA,MacLeod MK,Hale BG,Patakas A,Garside P,Brewer JM
25-9985 was used in Flow cytometry/Cell sorting to investigate the conditions affecting follicular helper T cell differentiation and antigen presentation in an influenza infection model.
Mon Aug 10 00:00:00 EDT 2015
CD1d-dependent expansion of NKT follicular helper cells in vivo and in vitro is a product of cellular proliferation and differentiation.
International immunology
Rampuria P,Lang ML
25-9985 was used in Flow cytometry/Cell sorting to demonstrate that α-GC-stimulated, CD1d-dependent increase in peripheral NKT follicular helper cells is a result of cellular proliferation and differentiation.
Fri May 01 00:00:00 EDT 2015
Programmed cell death receptor ligand 1 modulates the regulatory T cells' capacity to repress shock/sepsis-induced indirect acute lung injury by recruiting phosphatase SRC homology region 2 domain-containing phosphatase 1.
Shock (Augusta, Ga.)
Tang L,Bai J,Chung CS,Lomas-Neira J,Chen Y,Huang X,Ayala A
25-9985 was used in Flow cytometry/Cell sorting to investigate the mechanisms by which the programmed cell death receptor 1 ligand suppresses lung injury.
Thu Jan 01 00:00:00 EST 2015
Active players in resolution of shock/sepsis induced indirect lung injury: immunomodulatory effects of Tregs and PD-1.
Journal of leukocyte biology
Tang L,Bai J,Chung CS,Lomas-Neira J,Chen Y,Huang X,Ayala A
25-9985 was used in Flow cytometry/Cell sorting to explore the ability of Tregs to modify the innate immune response induced by experimental iALI.
Sat Nov 01 00:00:00 EDT 2014
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003