Product References
NCR negative group 3 innate lymphoid cell (NCR- ILC3) participates in abnormal pathology of lung in cigarette smoking-induced COPD mice.
Immunity, inflammation and disease
Chu S,Ma L,Yang X,Xiao B,Liang Y,Zheng S,Li L
25-3351-82 was used in Flow cytometry/Cell sorting to investigate the role of Natural Cytotoxicity Receptor negative innate lymphoid cells (NCR- ILC3) in the pathogenesis of chronic obstructive pulmonary disease (COPD) induced by cigarette smoking.
Wed Mar 01 00:00:00 EST 2023
RIPK3 controls MAIT cell accumulation during development but not during infection.
Cell death & disease
Patton T,Zhao Z,Lim XY,Eddy E,Wang H,Nelson AG,Ennis B,Eckle SBG,Souter MNT,Pediongco TJ,Koay HF,Zhang JG,Djajawi TM,Louis C,Lalaoui N,Jacquelot N,Lew AM,Pellicci DG,McCluskey J,Zhan Y,Chen Z,Lawlor KE,Corbett AJ
25-3351-82 was used in Flow cytometry/Cell sorting to suggest that, distinct from conventional T cells, the accumulation of MAIT cells is restrained by RIPK3 signalling, likely prior to thymic egress, in a manner independent of canonical apoptotic and necroptotic cell death pathways.
Sat Feb 11 00:00:00 EST 2023
CD169-positive macrophages enhance abscopal effect of radiofrequency ablation therapy in liver cancer.
Translational oncology
Song X,Li N,Liu Y,Wang Z,Wang T,Tan S,Li C,Qiu C,Gao L,Asano K,Tanaka M,Liang X,Liu X,Ma C
25-3351-82 was used in Flow Cytometry to establish the mouse model mimicking radiofrequency ablation (RFA)-induced abscopal effect, in which RFA eliminated the local orthotopic liver tumor but failed to control growth of distant tumor.
Sat Jan 01 00:00:00 EST 2022
SOCS2 regulation of growth hormone signaling requires a canonical interaction with phosphotyrosine.
Bioscience reports
Li K,Meza Guzman LG,Whitehead L,Leong E,Kueh A,Alexander WS,Kershaw NJ,Babon JJ,Doggett K,Nicholson SE
25-3351-82 was used in Flow cytometry/Cell sorting to indicate the critical role of phosphotyrosine binding in SOCS2 function.
Thu Dec 22 00:00:00 EST 2022
NK cells in hypoxic skin mediate a trade-off between wound healing and antibacterial defence.
Nature communications
Sobecki M,Krzywinska E,Nagarajan S,Audigé A,Hu¿nh K,Zacharjasz J,Debbache J,Kerdiles Y,Gotthardt D,Takeda N,Fandrey J,Sommer L,Sexl V,Stockmann C
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Wed Aug 04 00:00:00 EDT 2021
Interferon Lambda Regulates Cellular and Humoral Immunity in Pristane-Induced Lupus.
International journal of molecular sciences
Aschman T,Schaffer S,Biniaris Georgallis SI,Triantafyllopoulou A,Staeheli P,Voll RE
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Fri Oct 29 00:00:00 EDT 2021
Flow cytometry-based ex vivo murine NK cell cytotoxicity assay.
STAR protocols
Wong P,Wagner JA,Berrien-Elliott MM,Schappe T,Fehniger TA
25-3351-82 was used in Flow Cytometry to measure in vivo activated murine NK cells' ability to kill target cells ex vivo.
Fri Mar 19 00:00:00 EDT 2021
Flow cytometry-based ex vivo murine NK cell cytotoxicity assay.
STAR protocols
Wong P,Wagner JA,Berrien-Elliott MM,Schappe T,Fehniger TA
25-3351-82 was used in Flow Cytometry to measure in vivo activated murine NK cells' ability to kill target cells ex vivo.
Fri Mar 19 00:00:00 EDT 2021
Th2 cells promote eosinophil-independent pathology in a murine model of allergic bronchopulmonary aspergillosis.
European journal of immunology
Dietschmann A,Schruefer S,Krappmann S,Voehringer D
25-3351-82 was used in Flow Cytometry to use a mouse model of ABPA to investigate the role of eosinophils and T cell-derived IL-4/IL-13 for induction of allergic lung inflammation.
Wed Jul 01 00:00:00 EDT 2020
Characterization of the pathoimmunology of necrotizing enterocolitis reveals novel therapeutic opportunities.
Nature communications
Cho SX,Rudloff I,Lao JC,Pang MA,Goldberg R,Bui CB,McLean CA,Stock M,Klassert TE,Slevogt H,Mangan NE,Cheng W,Fischer D,Gfroerer S,Sandhu MK,Ngo D,Bujotzek A,Lariviere L,Schumacher F,Tiefenthaler G,Beker F,Collins C,Kamlin COF,König K,Malhotra A,Tan K,Theda C,Veldman A,Ellisdon AM,Whisstock JC,Berger PJ,Nold-Petry CA,Nold MF
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Fri Nov 13 00:00:00 EST 2020
Hhex Directly Represses BIM-Dependent Apoptosis to Promote NK Cell Development and Maintenance.
Cell reports
Goh W,Scheer S,Jackson JT,Hediyeh-Zadeh S,Delconte RB,Schuster IS,Andoniou CE,Rautela J,Degli-Esposti MA,Davis MJ,McCormack MP,Nutt SL,Huntington ND
25-3351 was used in Flow cytometry/Cell sorting to show homeobox transcriptional regulator Hhex regulates NK cell homeostasis and immunity.
Tue Oct 20 00:00:00 EDT 2020
Stage-Specific Requirement for Eomes in Mature NK Cell Homeostasis and Cytotoxicity.
Cell reports
Wagner JA,Wong P,Schappe T,Berrien-Elliott MM,Cubitt C,Jaeger N,Lee M,Keppel CR,Marin ND,Foltz JA,Marsala L,Neal CC,Sullivan RP,Schneider SE,Keppel MP,Saucier N,Cooper MA,Fehniger TA
25-3351 was used in Flow cytometry/Cell sorting to develop a tamoxifen-inducible, type-1-ILC-specific (Ncr1-targeted) cre mouse and combine this with Eomes-floxed mice.
Tue Jun 02 00:00:00 EDT 2020
Novel CD11b+Gr-1+Sca-1+ myeloid cells drive mortality in bacterial infection.
Science advances
Park MY,Kim HS,Lee HY,Zabel BA,Bae YS
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Cancer-Specific Loss of p53 Leads to a Modulation of Myeloid and T Cell Responses.
Cell reports
Blagih J,Zani F,Chakravarty P,Hennequart M,Pilley S,Hobor S,Hock AK,Walton JB,Morton JP,Gronroos E,Mason S,Yang M,McNeish I,Swanton C,Blyth K,Vousden KH
25-3351 was used in Flow cytometry/Cell sorting to show suppressive regulatory T cells accumulate in tumour cells in the absence of p53 indicating a pathway in which cancer cells circumvent immune destruction.
Tue Jan 14 00:00:00 EST 2020
Synergized regulation of NK cell education by NKG2A and specific Ly49 family members.
Nature communications
Zhang X,Feng J,Chen S,Yang H,Dong Z
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2019
SCGN deficiency results in colitis susceptibility.
eLife
Sifuentes-Dominguez LF,Li H,Llano E,Liu Z,Singla A,Patel AS,Kathania M,Khoury A,Norris N,Rios JJ,Starokadomskyy P,Park JY,Gopal P,Liu Q,Tan S,Chan L,Ross T,Harrison S,Venuprasad K,Baker LA,Jia D,Burstein E
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Wed Oct 30 00:00:00 EDT 2019
LIF regulates CXCL9 in tumor-associated macrophages and prevents CD8+ T cell tumor-infiltration impairing anti-PD1 therapy.
Nature communications
Pascual-García M,Bonfill-Teixidor E,Planas-Rigol E,Rubio-Perez C,Iurlaro R,Arias A,Cuartas I,Sala-Hojman A,Escudero L,Martínez-Ricarte F,Huber-Ruano I,Nuciforo P,Pedrosa L,Marques C,Braña I,Garralda E,Vieito M,Squatrito M,Pineda E,Graus F,Espejo C,Sahuquillo J,Tabernero J,Seoane J
25-3351-82 was used in Flow Cytometry to study the effect of the combination of LIF neutralising antibodies with the inhibition of the PD1 immune checkpoint on tumour progression.
Tue Jun 11 00:00:00 EDT 2019
E3 Ligase VHL Promotes Group 2 Innate Lymphoid Cell Maturation and Function via Glycolysis Inhibition and Induction of Interleukin-33 Receptor.
Immunity
Li Q,Li D,Zhang X,Wan Q,Zhang W,Zheng M,Zou L,Elly C,Lee JH,Liu YC
25-3351 was used in Flow Cytometry to indicate that the VHL-HIF-glycolysis axis is essential for the late-stage maturation and function of ILC2s via targeting IL-33-ST2 pathway.
Tue Feb 20 00:00:00 EST 2018
Regulation of Intracellular Triiodothyronine Is Essential for Optimal Macrophage Function.
Endocrinology
van der Spek AH,Surovtseva OV,Jim KK,van Oudenaren A,Brouwer MC,Vandenbroucke-Grauls CMJE,Leenen PJM,van de Beek D,Hernandez A,Fliers E,Boelen A
25-3351 was used in Flow cytometry/Cell sorting to study the role of optimal intracellular concentrations of the active thyroid hormone triiodothyronine in proinflammatory macrophage function.
Tue May 01 00:00:00 EDT 2018
The Thyroid Hormone Inactivating Type 3 Deiodinase Is Essential for Optimal Neutrophil Function: Observations From Three Species.
Endocrinology
van der Spek AH,Jim KK,Karaczyn A,van Beeren HC,Ackermans MT,Darras VM,Vandenbroucke-Grauls CMJE,Hernandez A,Brouwer MC,Fliers E,van de Beek D,Boelen A
25-3351 was used in Flow cytometry/Cell sorting to study whether type 3 deiodinase plays a role in neutrophil function during infection.
Thu Feb 01 00:00:00 EST 2018
Conventional NK cells and ILC1 are partially ablated in the livers of Ncr1 iCreTbx21 fl/fl mice.
Wellcome open research
Cuff AO,Male V
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Sat Jan 09 00:00:00 EST 2021
Loss of HIF-1α in natural killer cells inhibits tumour growth by stimulating non-productive angiogenesis.
Nature communications
Krzywinska E,Kantari-Mimoun C,Kerdiles Y,Sobecki M,Isagawa T,Gotthardt D,Castells M,Haubold J,Millien C,Viel T,Tavitian B,Takeda N,Fandrey J,Vivier E,Sexl V,Stockmann C
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Fri Nov 17 00:00:00 EST 2017
Characterisation of innate lymphoid cell populations at different sites in mice with defective T cell immunity.
Wellcome open research
Dutton EE,Camelo A,Sleeman M,Herbst R,Carlesso G,Belz GT,Withers DR
25-3351-82 was used in Flow Cytometry to compare the innate lymphoid cell populations at different sites in mice with defective T cell immunity.
Sat Jan 09 00:00:00 EST 2021
Boosting the hypoxic response in myeloid cells accelerates resolution of fibrosis and regeneration of the liver in mice.
Oncotarget
Kantari-Mimoun C,Krzywinska E,Castells M,Milien C,Klose R,Meinecke AK,Lemberger U,Mathivet T,Gojkovic M,Schrödter K,Österreicher C,Fandrey J,Rundqvist H,Stockmann C
25-3351 was used in Flow cytometry/Cell sorting to show that the resolution of fibrosis can be accelerated by deleting VHL, a negative regulator of HIFs and VEGF, specifically in myeloid cells in mice.
Tue Feb 28 00:00:00 EST 2017
The breast tumor microenvironment alters the phenotype and function of natural killer cells.
Cellular & molecular immunology
Krneta T,Gillgrass A,Chew M,Ashkar AA
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
NK cell development requires Tsc1-dependent negative regulation of IL-15-triggered mTORC1 activation.
Nature communications
Yang M,Chen S,Du J,He J,Wang Y,Li Z,Liu G,Peng W,Zeng X,Li D,Xu P,Guo W,Chang Z,Wang S,Tian Z,Dong Z
Published figure using CD335 (NKp46) monoclonal antibody (Product # 25-3351-82) in Flow Cytometry
Wed Sep 07 00:00:00 EDT 2016
Targeting VEGF-A in myeloid cells enhances natural killer cell responses to chemotherapy and ameliorates cachexia.
Nature communications
Klose R,Krzywinska E,Castells M,Gotthardt D,Putz EM,Kantari-Mimoun C,Chikdene N,Meinecke AK,Schrödter K,Helfrich I,Fandrey J,Sexl V,Stockmann C
25-3351 was used in Flow cytometry/Cell sorting to investigate the effect of VEGF-A genetic inactivation in myeloid cells on natural killer cell responses and chemotherapy-induced cachexia.
Fri Aug 19 00:00:00 EDT 2016
Interferon-γ from Brain Leukocytes Enhances Meningitis by Type 4 Streptococcus pneumoniae.
Frontiers in microbiology
Pettini E,Fiorino F,Cuppone AM,Iannelli F,Medaglini D,Pozzi G
25-3351 was used in Flow cytometry/Cell sorting to elucidate the role of IFN-γ in the neuroinflammatory response following type 4 Streptococcus pneumoniae-induced meningitis in mice.
Wed Dec 09 00:00:00 EST 2015
IL-22 bridges the lymphotoxin pathway with the maintenance of colonic lymphoid structures during infection with Citrobacter rodentium.
Nature immunology
Ota N,Wong K,Valdez PA,Zheng Y,Crellin NK,Diehl L,Ouyang W
Sun Aug 28 00:00:00 EDT 2011