Product References
ADAM12 is a costimulatory molecule that determines Th1 cell fate and mediates tissue inflammation.
Cellular & molecular immunology
Liu Y,Bockermann R,Hadi M,Safari I,Carrion B,Kveiborg M,Issazadeh-Navikas S
16-7041-81 was used in Flow Cytometry to study the role of ADAM12 in T-cell responses, fate determination on activation, and its functions in T cells to mediate tissue inflammation.
Sun Aug 01 00:00:00 EDT 2021
Activation of regulatory T cells triggers specific changes in glycosylation associated with Siglec-1-dependent inflammatory responses.
Wellcome open research
Wu G,Murugesan G,Nagala M,McCraw A,Haslam SM,Dell A,Crocker PR
16-7041-81 was used in Cell Culture to investigate the molecular basis for Siglec-1 binding to regulatory T cells using in vitro-induced cells as a model system.
Tue Mar 01 00:00:00 EST 2022
Inhibition of Glycolysis in Pathogenic TH17 Cells through Targeting a miR -21-Peli1-c-Rel Pathway Prevents Autoimmunity.
Journal of immunology (Baltimore, Md. : 1950)
Qiu R,Yu X,Wang L,Han Z,Yao C,Cui Y,Hou G,Dai D,Jin W,Shen N
16-7041-81 was used in Functional assay to investigate if CNS-infiltrated pathogenic TH17 cells from diseased mice specifically upregulate glycolytic pathway genes compared with homeostatic intestinal TH17 cells.
Mon Jun 15 00:00:00 EDT 2020
MiR-1165-3p Suppresses Th2 Differentiation via Targeting IL-13 and PPM1A in a Mouse Model of Allergic Airway Inflammation.
Allergy, asthma & immunology research
Wang Z,Ji N,Chen Z,Sun Z,Wu C,Yu W,Hu F,Huang M,Zhang M
16-7041-95 was used in T-Cell Activation to investigate the associations between Th2 differentiation and miR-1165b-3p in asthma as well as the possible mechanisms.
Tue Sep 01 00:00:00 EDT 2020
The MicroRNA miR-22 Represses Th17 Cell Pathogenicity by Targeting PTEN-Regulated Pathways.
ImmunoHorizons
Wang L,Qiu R,Zhang Z,Han Z,Yao C,Hou G,Dai D,Jin W,Tang Y,Yu X,Shen N
16-7041-95 was used in In vitro Assay to identify that Th17 cell-intrinsic miR-22 could protect mice from autoimmunity by targeting PTEN-regulated pathways.
Tue Jun 09 00:00:00 EDT 2020
Runx-mediated regulation of CCL5 via antagonizing two enhancers influences immune cell function and anti-tumor immunity.
Nature communications
Seo W,Shimizu K,Kojo S,Okeke A,Kohwi-Shigematsu T,Fujii SI,Taniuchi I
16-7041-81 was used in ELISA to suggest that RUNX3-mediated CCL5 repression is critical for modulating anti-tumor immunity.
Thu Mar 26 00:00:00 EDT 2020
Gasdermin D in peripheral myeloid cells drives neuroinflammation in experimental autoimmune encephalomyelitis.
The Journal of experimental medicine
Li S,Wu Y,Yang D,Wu C,Ma C,Liu X,Moynagh PN,Wang B,Hu G,Yang S
16-7041-81 was used in Flow Cytometry to further define the role of gasdermin D in experimental autoimmune encephalomyelitis.
Mon Nov 04 00:00:00 EST 2019
Characterization of articular cartilage homeostasis and the mechanism of superior cartilage regeneration of MRL/MpJ mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Deng Z,Gao X,Sun X,Amra S,Lu A,Cui Y,Eltzschig HK,Lei G,Huard J
16-7041 was used in Immunohistochemistry-immunofluorescence to investigate articular cartilage (AC) homeostasis and signaling pathways involved in the superior cartilage regeneration of Murphy Roths large (MRL/MpJ) mice.
Thu Aug 01 00:00:00 EDT 2019
Fetal T Cell Activation in the Amniotic Cavity during Preterm Labor: A Potential Mechanism for a Subset of Idiopathic Preterm Birth.
Journal of immunology (Baltimore, Md. : 1950)
Gomez-Lopez N,Romero R,Xu Y,Miller D,Arenas-Hernandez M,Garcia-Flores V,Panaitescu B,Galaz J,Hsu CD,Para R,Berry SM
16-7041-95 was used in T-Cell Activation to show that fetal T cells are a predominant leukocyte population in amniotic fluid during preterm gestrations, and that chord blood T cells obtained from women with idiopathic preterm labor and birth displayed enhanced ex vivo activation.
Tue Oct 01 00:00:00 EDT 2019
Proteomic Profiling of Mouse Helper T Cell Differentiation.
Proteomics
van den Ham HJ,Binai NA,Zaaraoui-Boutahar F,Heck AJR,Andeweg AC
16-7041-81 was used in In vitro Assay to support the design of prophylactic and therapeutic intervention strategies for (infectious) diseases.
Mon Apr 01 00:00:00 EDT 2019
The Sympathetic Nervous System Mitigates CNS Autoimmunity via β2-Adrenergic Receptor Signaling in Immune Cells.
Cell reports
Araujo LP,Maricato JT,Guereschi MG,Takenaka MC,Nascimento VM,de Melo FM,Quintana FJ,Brum PC,Basso AS
16-7041-81 was used in Flow Cytometry to uncover a suppressive role for the SNS in CNS autoimmunity while they identify potential targets for therapeutic intervention.
Tue Sep 17 00:00:00 EDT 2019
Distinct Regulation of Th17 and Th1 Cell Differentiation by Glutaminase-Dependent Metabolism.
Cell
Johnson MO,Wolf MM,Madden MZ,Andrejeva G,Sugiura A,Contreras DC,Maseda D,Liberti MV,Paz K,Kishton RJ,Johnson ME,de Cubas AA,Wu P,Li G,Zhang Y,Newcomb DC,Wells AD,Restifo NP,Rathmell WK,Locasale JW,Davila ML,Blazar BR,Rathmell JC
16-7041-81 was used in Functional assays to identify a key role for glutaminase (GLS) in T cell activation and specification.
Thu Dec 13 00:00:00 EST 2018
MicroRNA-210 overexpression promotes psoriasis-like inflammation by inducing Th1 and Th17 cell differentiation.
The Journal of clinical investigation
Wu R,Zeng J,Yuan J,Deng X,Huang Y,Chen L,Zhang P,Feng H,Liu Z,Wang Z,Gao X,Wu H,Wang H,Su Y,Zhao M,Lu Q
16-7041-81 was used in In vitro experiments to study the role of microRNA-210 in the immune imbalance of T lymphocyte subsets in psoriasis and suggest a potential therapeutic avenue.
Fri Jun 01 00:00:00 EDT 2018
Cytochrome c Oxidase Activity Is a Metabolic Checkpoint that Regulates Cell Fate Decisions During T Cell Activation and Differentiation.
Cell metabolism
Tarasenko TN,Pacheco SE,Koenig MK,Gomez-Rodriguez J,Kapnick SM,Diaz F,Zerfas PM,Barca E,Sudderth J,DeBerardinis RJ,Covian R,Balaban RS,DiMauro S,McGuire PJ
16-7041-81 was used in Flow cytometry/Cell sorting to understand the mechanism of cytochrome c oxidase regulation of OXPHOS using a mouse model with isolated dysfunction in T cells by targeting the COX10 gene which produces mitochondrial disease in humans.
Tue Jun 06 00:00:00 EDT 2017
Paeoniflorin Ameliorates Experimental Autoimmune Encephalomyelitis via Inhibition of Dendritic Cell Function and Th17 Cell Differentiation.
Scientific reports
Zhang H,Qi Y,Yuan Y,Cai L,Xu H,Zhang L,Su B,Nie H
16-7041 was used in Flow cytometry/Cell sorting to investigate the effect Paeoniflorin has on Th cells and dendritic cells in a mouse MS model.
Mon Feb 06 00:00:00 EST 2017
The RNA-binding protein HuR contributes to neuroinflammation by promoting C-C chemokine receptor 6 (CCR6) expression on Th17 cells.
The Journal of biological chemistry
Chen J,Martindale JL,Cramer C,Gorospe M,Atasoy U,Drew PD,Yu S
16-7041 was used in T-Cell Activation experiments to study how HuR contributes to T helper 17 cell-mediated autoimmune neuroinflammation, and its potential therapeutic target for managing autoimmune conditions of the central nervous system.
Fri Sep 01 00:00:00 EDT 2017
Metabolic control of TH17 and induced Treg cell balance by an epigenetic mechanism.
Nature
Xu T,Stewart KM,Wang X,Liu K,Xie M,Ryu JK,Li K,Ma T,Wang H,Ni L,Zhu S,Cao N,Zhu D,Zhang Y,Akassoglou K,Dong C,Driggers EM,Ding S
16-7041-81 was used in Immunohistochemistry, ELISA, Functional Assay to investigate T helper 17 cell differentiation and fate.
Thu Aug 10 00:00:00 EDT 2017
IL-15-dependent balance between Foxp3 and RORγt expression impacts inflammatory bowel disease.
Nature communications
Tosiek MJ,Fiette L,El Daker S,Eberl G,Freitas AA
16-7041 was used in an ELISA assay to demonstrate that impaired delivery of IL-15 to CD4+ T cells in the colon down-modulates Foxp3 expression and enhances RORγt expression.
Fri Mar 11 00:00:00 EST 2016
NADPH Oxidases Are Essential for Macrophage Differentiation.
The Journal of biological chemistry
Xu Q,Choksi S,Qu J,Jang J,Choe M,Banfi B,Engelhardt JF,Liu ZG
16-7041 was used in Functional assays to investigate the role of NOX1 and NOX2 in macrophage differentiation and tumor progression.
Fri Sep 16 00:00:00 EDT 2016
Recombinant adeno-vaccine expressing enterovirus 71-like particles against hand, foot, and mouth disease.
PLoS neglected tropical diseases
Tsou YL,Lin YW,Shao HY,Yu SL,Wu SR,Lin HY,Liu CC,Huang C,Chong P,Chow YH
16-7041 was used in an ELISpot assay to develop a recombinant adenovirus Ad-EVVLP for the treatment of hand, foot and mouth disease, providing a possible vaccine for future immunisation against the disease.
Wed Apr 01 00:00:00 EDT 2015
Basophils regulate the recruitment of eosinophils in a murine model of irritant contact dermatitis.
The Journal of allergy and clinical immunology
Nakashima C,Otsuka A,Kitoh A,Honda T,Egawa G,Nakajima S,Nakamizo S,Arita M,Kubo M,Miyachi Y,Kabashima K
16-7041 was used in Inhibition experiments to clarify the roles of eosinophils in irritant contact dermatitis and the underlying mechanism of eosinophil infiltration of the skin.
Tue Jul 01 00:00:00 EDT 2014
In vitro generated Th17 cells support the expansion and phenotypic stability of CD4(+)Foxp3(+) regulatory T cells in vivo.
Cytokine
Zhou Q,Hu Y,Howard OM,Oppenheim JJ,Chen X
16-7041 was used in Functional assays to demonstrate that the TNF-TNFR2 pathway plays a crucial role in the reciprocal stimulatory effect of Th17 cells and Tregs.
Wed Jan 01 00:00:00 EST 2014
Single-cell RNA sequencing reveals T helper cells synthesizing steroids de novo to contribute to immune homeostasis.
Cell reports
Mahata B,Zhang X,Kolodziejczyk AA,Proserpio V,Haim-Vilmovsky L,Taylor AE,Hebenstreit D,Dingler FA,Moignard V,Göttgens B,Arlt W,McKenzie AN,Teichmann SA
16-7041 was used in Functional assays to demonstrate the production of the steroid pregnenolone by Th2 cells in vitro and in vivo in a helminth infection model.
Thu May 22 00:00:00 EDT 2014
Staphylococcus aureus phenol-soluble modulin peptides modulate dendritic cell functions and increase in vitro priming of regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Schreiner J,Kretschmer D,Klenk J,Otto M,Bühring HJ,Stevanovic S,Wang JM,Beer-Hammer S,Peschel A,Autenrieth SE
16-7041 was used in Functional assays to demonstrate that PSMs from highly virulent pathogens influence dendritic cell functions, thereby modulating the adaptive immune response.
Mon Apr 01 00:00:00 EDT 2013
Recent thymic emigrants are biased against the T-helper type 1 and toward the T-helper type 2 effector lineage.
Blood
Hendricks DW,Fink PJ
16-7041 was used in Flow cytometry/Cell sorting to investigate the relationships between recent thymic emigrants and T-helper type 1 and 2 cells.
Thu Jan 27 00:00:00 EST 2011
Soluble flagellin, FliC, induces an Ag-specific Th2 response, yet promotes T-bet-regulated Th1 clearance of Salmonella typhimurium infection.
European journal of immunology
Bobat S,Flores-Langarica A,Hitchcock J,Marshall JL,Kingsley RA,Goodall M,Gil-Cruz C,Serre K,Leyton DL,Letran SE,Gaspal F,Chester R,Chamberlain JL,Dougan G,López-Macías C,Henderson IR,Alexander J,MacLennan IC,Cunningham AF
16-7041 was used in an ELISPOT assay to show that, in mice, immunisation with soluble, recombinant FliC protein flagellin induces Th2 responses.
Wed Jun 01 00:00:00 EDT 2011
The metalloprotease-disintegrin ADAM8 is essential for the development of experimental asthma.
American journal of respiratory and critical care medicine
Naus S,Blanchet MR,Gossens K,Zaph C,Bartsch JW,McNagny KM,Ziltener HJ
16-7041 was used in Flow cytometry/Cell sorting to determine that ADAM8 plays a crucial proinflammatory role in airway inflammation.
Tue Jun 15 00:00:00 EDT 2010
Potent anti-tumor responses to immunization with dendritic cells loaded with tumor tissue and an NKT cell ligand.
Immunology and cell biology
Petersen TR,Sika-Paotonu D,Knight DA,Dickgreber N,Farrand KJ,Ronchese F,Hermans IF
16-7041 was used in Flow cytometry/Cell sorting to investigate the impact of eliciting T-cell help from activated iNKT cells using alpha-galactosylceramide, showing potent anti-tumour response.
Thu Jul 01 00:00:00 EDT 2010
Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Journal of immunology (Baltimore, Md. : 1950)
Weber KS,Hildner K,Murphy KM,Allen PM
16-7041 was used in Functional assays to study the regulation of unique Th cell subset calcium signals and their correlation to biological functions, showing that Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Wed Sep 01 00:00:00 EDT 2010
Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Journal of immunology (Baltimore, Md. : 1950)
Weber KS,Hildner K,Murphy KM,Allen PM
16-7041 was used in Functional assays to study the regulation of unique Th cell subset calcium signals and their correlation to biological functions, showing that Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Wed Sep 01 00:00:00 EDT 2010
Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Journal of immunology (Baltimore, Md. : 1950)
Weber KS,Hildner K,Murphy KM,Allen PM
16-7041 was used in Functional assays to study the regulation of unique Th cell subset calcium signals and their correlation to biological functions, showing that Trpm4 differentially regulates Th1 and Th2 function by altering calcium signaling and NFAT localization.
Wed Sep 01 00:00:00 EDT 2010
Epigenetic regulation of Foxp3 expression in regulatory T cells by DNA methylation.
Journal of immunology (Baltimore, Md. : 1950)
Lal G,Zhang N,van der Touw W,Ding Y,Ju W,Bottinger EP,Reid SP,Levy DE,Bromberg JS
16-7041 was used in Functional assays to investigate Foxp3 as the master switch for Tregs, showing that expression is regulated by DNA methylation in Tregs.
Thu Jan 01 00:00:00 EST 2009
STAT6 deletion converts the Th2 inflammatory pathology afflicting Lat(Y136F) mice into a lymphoproliferative disorder involving Th1 and CD8 effector T cells.
Journal of immunology (Baltimore, Md. : 1950)
Archambaud C,Sansoni A,Mingueneau M,Devilard E,Delsol G,Malissen B,Malissen M
16-7041 was used in Flow cytometry/Cell sorting to study the effect of STAT6 delection on Lat(Y136F) mice.
Sun Mar 01 00:00:00 EST 2009