Search Thermo Fisher Scientific
Because bovine mastitis can cause significant losses, farmers and herd managers need quick diagnostic results so they can take fast and appropriate action. Traditionally, culture has been used for the diagnosis of mastitis infections, but results can take a long time and pathogens can be difficult to grow, especially for Mycoplasma species.
Our multiplex qPCR and sample extraction solutions, the Applied Biosystems VetMAX MastiType multiplex qPCR kits, together with the Applied Biosystems MagMAX CORE Nucleic Acid Purification Kit, are designed to provide same-day results even for Mycoplasma spp., enabling farmers and veterinarians to take immediate and well-informed action.
VetMAX MastiType and PathoProof products belong to the same family of multiplex qPCRs for the detection of mastitis-causing pathogens in milk. To consolidate our portfolio for mastitis diagnostics, PathoProof products are now available through our Animal Health team.
Your advantages:
The VetMAX MastiType Micro4 Kit offers real-time PCR detection of mastitis-causing pathogens in a single-well real-time PCR reaction. The VetMAX MastiType Micro4 assay enables rapid, accurate detection of the major contagious mastitis-causing pathogens in bovine milk using real-time PCR amplification of DNA unique to each pathogen.
Regulatory requirements vary by country; products may not be available in your geographic area.
VetMAX MastiType Myco8 Kit offers real‑time PCR detection of eight mastitis-causing pathogens in two separate PCR reactions. The VetMAX MastiType Myco8 assay enables rapid, accurate detection of mastitis-causing pathogens, especially the highly contagious and difficult-to-culture mycoplasma strains in bovine milk, using real-time PCR amplification of DNA unique to each pathogen.
Regulatory requirements vary by country; products may not be available in your geographic area.
The VetMAX MastiType Multi Kit offers real-time PCR detection of all major mastitis-causing pathogens and a penicillin-resistance gene in four separate PCR reactions. The VetMAX MastiType Multi assay helps to enable rapid, accurate detection of 15 major mastitis-causing pathogens in bovine milk using real-time PCR amplification of DNA unique to each pathogen.
The VetMAX MastiType Micro4 Kit offers real-time PCR detection of mastitis-causing pathogens in a single-well real-time PCR reaction. The VetMAX MastiType Micro4 assay enables rapid, accurate detection of the major contagious mastitis-causing pathogens in bovine milk using real-time PCR amplification of DNA unique to each pathogen.
Regulatory requirements vary by country; products may not be available in your geographic area.
VetMAX MastiType Myco8 Kit offers real‑time PCR detection of eight mastitis-causing pathogens in two separate PCR reactions. The VetMAX MastiType Myco8 assay enables rapid, accurate detection of mastitis-causing pathogens, especially the highly contagious and difficult-to-culture mycoplasma strains in bovine milk, using real-time PCR amplification of DNA unique to each pathogen.
Regulatory requirements vary by country; products may not be available in your geographic area.
The VetMAX MastiType Multi Kit offers real-time PCR detection of all major mastitis-causing pathogens and a penicillin-resistance gene in four separate PCR reactions. The VetMAX MastiType Multi assay helps to enable rapid, accurate detection of 15 major mastitis-causing pathogens in bovine milk using real-time PCR amplification of DNA unique to each pathogen.
The VetMAX MastiType multiplex qPCR kits allow simultaneous diagnosis of mastitis-causing pathogens, enabling labs to provide farmers and veterinarians with results to take immediate and informed action.
Simplify, accelerate, and standardize data analysis with our result interpretation software. The cloud-based Animal Health VeriVet App allows you to create a plate layout, generate a template file for import to the real-time PCR instrument, and remotely monitor the instrument run.
Bovine mastitis is the most common and costly disease among dairy cows. It is an inflammation of the mammary gland that is predominantly caused by a bacterial infection. When mastitis is identified in a cow’s quarter(s), it is important to identify the pathogen causing the infection because different categories of pathogens require different management strategies. Once the infection is identified, a dairy farmer can work with his or her veterinarian to target an effective treatment plan. Prudent use of antibiotics reduces the likelihood of resistant pathogens developing and can reduce the duration of treatment a cow may need, which in turn decreases operation costs.
Mastitis is estimated to cost the global dairy industry US $19.7 to US $32 billion annually.
With clinical mastitis, milk is typically visibly watery, thick or ropy, flakey, flocky, containing pus and blood or clots and often the quarter is red, swollen, and painful. A case of clinical mastitis may lead to an economic loss that ranges somewhere between US $128 (€105) and US $444 ( €360) (Rollin et al. 2015; Cha et al. 2011; Huijps et al. 2008)
Subclinical mastitis is not visibly notable when looking at the milk, and the udder appears normal. However, the somatic cell count of the cow’s milk is slightly elevated, and microorganisms can usually be detected in a diagnostic sample. The overall product loss of subclinical mastitis is estimated at US $110 per cow annually. The greatest economic losses are attributed to high bulk tank somatic cell count resulting in loss of quality premiums and overall lower milk production.
The cost of M. bovis in cattle due to mastitis is estimated at US $108 million annually with infection rates of up to 70% of a herd, for pneumonia and related respiratory illnesses at US $32 million annually. Total M. bovis estimated disease cost is at least US $140 million annually in the U.S. Even higher losses—estimated at €144 to €192 million—have been reported in Europe.
Mastitis diagnosis utilizing a turnkey solution from Thermo Fisher Scientific’s Animal Health Group
Presented at the IDF mastitis conference 2019 in Copenhagen, Denmark
Fast and convenient nucleic acid extraction method for mastitis diagnostic
Presented at the IDF mastitis conference 2019 in Copenhagen, Denmark
Real-time PCR data analysis tool utilizing a novel cloud-based software for easy interpretation of animal pathogen detection
Presented at the IDF mastitis conference 2019 in Copenhagen, Denmark
To consolidate our portfolio for mastitis diagnostics, PathoProof products can now be ordered through the Animal Health.
PathoProof kits—Applied Biosystems 7500 / QS | |
---|---|
PF8100S | PathoProof Mycoplasma-8 (Applied Biosystems 7500) |
PF8100SB | qPCR reagents for use with PF8100S and PF8100SKF |
PF1650S | PathoProof Complete-16 |
PF1650SB | qPCR reagents for use with PF1650S and PF1650SKF |
PF1650L | PathoProof Complete-16 |
PF1650LKF | PathoProof Complete-16 KF |
PF1650LB | qPCR reagents for use with PF1650L and PF1650LKF |
PF3050S | PathoProof Major-3 |
PF3050SB | qPCR reagents for use with PF3050S and PF3050SKF |
PF3050L | PathoProof Major-3 |
PF3050LB | qPCR reagents for use with PF3050L and PF3050LKF |
PF4250L | PathoProof Major-4.2 |
PF4250LB | qPCR reagents for use with PF4250L and PF4250LKF |
PathoProof kits—Agilent | |
---|---|
PF0914S | PathoProof Mastitis Major-3 50 Tests |
PF0914SB | qPCR reagents for use with PF0914S |
PF0914L | PathoProof Mastitis Major-3 4x96 Tests |
PF0914LB | qPCR reagents for use with PF0914L |
PF8500S | PathoProof Mycoplasma-8 (Agilent qPCR) |
PF8500SB | qPCR reagents for use with PF8500S &PF8500SKF |
PF8500L | PathoProof Mycoplasma-8 (Agilent qPCR) |
PF8500LKF | PathoProof Mycoplasma-8 KF (Agilent qPCR) |
PF8500LB | qPCR reagents for use with PF8500L and PF8500LKF |
PF1600S | PathoProof Mastitis Complete-16 50 Tests |
PF1600SB | qPCR reagents for use with PF1600S |
PF1600L | PathoProof Mastitis Complete-16 4x96 |
PF1600LKF | PathoProof Mastitis Complete-16 KF 384 tests |
PF1600LB | qPCR reagents for use with PF1600L |
PF0942S | PathoProof Mastitis Major-4.2 50 Tests |
PF0942SB | qPCR reagents for use with PF0942S |
PF0942L | PathoProof Mastitis Major-4.2 4x96 Tests |
PF0942LB | qPCR reagents for use with PF0942L |
PathoProof instruments and software | |
---|---|
PF0888A | PathoProof Mastitis Software |
If you are an existing PathoProof customer, you can enter the SKU in our Quick Order tool.
For Veterinary Use Only. For In Vitro Use Only. Regulatory requirements vary by country; products may not be available in your geographic area.