B·R·A·H·M·S KRYPTOR Analysers
Uncompromising quality – supporting relevant clinical decisions
Explore B·R·A·H·M·S KRYPTOR analysers
A precise measurement platform using TRACE technology
For reliable results with biomarker assays, a precise measurement platform is necessary. The Thermo Scientific B·R·A·H·M·S KRYPTOR Analysers are trusted instruments that can perform various analyses in random-access operations. The fully automatic, closed laboratory analysis systems are exclusively licenced to use the Nobel-prize-winning measuring principle TRACE (Time Resolved Amplified Cryptate Emission).
With our excellent clinical and technological expertise, we strive to support our customers with products and services that go beyond expectations. We aim to enrich the quality of our customers’ work and enable clinicians to improve patient management.
  Informational: This product is not available for purchase by the general public
Improving patient management with B·R·A·H·M·S KRYPTOR analysers
Providing access to new innovative markers
Trusted results with outstanding precision
For trusted results, precision and accuracy are essential. Nobel Prize-winning TRACE Technology provides the highest precision possible for the entire measurement range.
Reliable decisions from low interference
Interferences will cause errors and result in inaccuracy. Low interference leads to safe decision-making when the laboratory interprets patient results providing the right course of treatment by a clinician. TRACE Technology and homogeneous immunoassay provide the lowest level of Human anti-mouse antibody or human anti-murine antibody (HAMA) interferences and low inter and intra-assay Coefficient of Variability (CV %).
Optimised workflow with self-determining dilution factor
Optimised workflows will enhance the time to result and reduce hands-on time. With no user intervention, TRACE Technology on B·R·A·H·M·S KRYPTOR Analysers can autonomously determine the dilution factor for out-of-range samples and can complete the dilution automatically.
Unique assays for essential indications
B·R·A·H·M·S KRYPTOR homogenous immunoassays are uniquely designed to fulfil routine laboratory needs with high-quality and reliable test menus. They are biotin-free, wash-free and separation free. Lot-to-lot stability provides the highest quality for essential indications.
Optimise workflow to focus on essentials
Fully automated maintenance – For reliable workflow
Ensure maintenance steps are not forgotten by using a predominantly automated and software-controlled maintenance management system. Track and record all maintenance steps to support the quality management system.
Real-time notification system – For keeping track of the samples and reagents
Get the results directly delivered via LIS connection. In addition to warning signals, get real-time notification when a bubble or particle is detected.
End-to-end traceability – For reducing error in data entry
Minimise errors by using barcoded reagents and consumables.
STAT function – For fast sample analysis during an emergency situation
Run emergency samples without interrupting the ongoing measurement.
User-friendly interface – For ease of use
React with purpose and ensure results are relevant when intuitive graphics like colour-coded system (“traffic light system”) alert run status and any necessary action.
Self-determining dilution factor – For no user intervention
Reduce the hands-on time using TRACE technology that helps determine the dilution factor for out-of-range samples and ensures that the dilution is carried out with no human intervention.
Choosing the right system for your lab
The B·R·A·H·M·S KRYPTOR analysers are designed to measure a range of biomarker assays, providing results for the following clinical areas:
  • Infectious disease:
    For the early detection of sepsis infection and improved patient prioritisation and prognosis in emergency and ICU.
  • Prenatal screening:
    For improved diagnosis of pre-eclampsia and chromosomal abnormalities.
B·R·A·H·M·S KRYPTOR GOLD
The Thermo Scientific™ B·R·A·H·M·S™ KRYPTOR™ GOLD is suited to medium size routine labs with up to 16 different immunoassays and with a throughput of up to approximately 115 tests per hour.
The novel engineering and thoughtful design of B·R·A·H·M·S KRYPTOR GOLD redefines user autonomy, throughput and cost efficiency.
A natural user experience - seamless interaction with B·R·A·H·M·S KRYPTOR GOLD guides calibration, processing and sample tracking regardless of user experience.
  • Intuitive interaction with simple, easy-to-understand icons on the main screen control all essential processes and simplify navigation
  • Purposeful visualisation with a colour-coded diagram of the instrument carousel communicates real-time run status and any necessary actions
  • Calibration and standard curves, as well as quality control results, are summarised graphically for efficient evaluation
  • The status window on the main screen displays the run status, reagent expiry date, upcoming maintenance, and specimen processing stage
  • Live lists of results and work to be done ensure processing clarity and allow entering new measurement requests at any time
B·R·A·H·M·S KRYPTOR compact PLUS
The Thermo Scientific B·R·A·H·M·S KRYPTOR compact PLUS is perfect for small-size routine labs or research labs with a requirement of up to 8 different immunoassays and a throughput of up to approximately 60 tests per hour.
With small footprint and intelligent engineering, Thermo Scientific B·R·A·H·M·S KRYPTOR compact PLUS brings exceptional value to clinical and research laboratories.
KRYPTOR Augmented Reality (AR)
Experience Thermo Scientific™ B·R·A·H·M·S™ KRYPTOR™ analysers in a new and exciting way.
  • Visualise B·R·A·H·M·S KRYPTOR analysers in your lab space
  • Discover all product highlights and special features
  • Explore the B·R·A·H·M·S Biomarkers test menu with carefully designed unique immunoassays
  • Learn more about the Nobel Prize-winning TRACE technology, exclusively available on B·R·A·H·M·S KRYPTOR analysers
  • Find out how laboratories benefit from Self-determining Dilution Factor (SDDF)
  • Understand how B·R·A·H·M·S KRYPTOR analysers will optimise your lab workflow
Resources
Video: B·R·A·H·M·S KRYPTOR analysers overview
The availability of products may vary in each country depending on local regulatory requirements.
Contact your local specialist for more details