Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a comprehensive range of products for your respiratory management needs. Our respiratory support products allows for the provision of supplemental oxygen, and our airway management products are designed for the safe and effective management of airways and containment of fluids. In addition, our Busse mucus specimen trap products prevent tube separation and contamination.
Our range of oxygen, aerosol, and anaesthetic masks are made from high quality materials. Adult and paediatric masks feature adjustable nose clips for custom fitting and an adjustable elastic braid strap. All masks are compatible with standard tubing connectors.
Our oxygen tubing products are manufactured from flexible non-toxic material. They are available in both standard or non-crush lumen varieties. The aerosol corrugated tubing is flexible and lightweight for maximum performance. These are available in a variety of lengths with cuttable cuffs every 15 cm for convenient length adjustments.
Our nasal cannulae are available in two different styles: Soft-Touch and Standard. The soft-touch Nasal Cannula products are made using a special manufacturing process which results in a flexible soft finish for increased patient comfort. They are lightweight, fit comfortably over the ear, and are available with straight tips for infants and flared curved tips in adult and paediatric sizes. Standard Cannula are available in a variety of configurations which include oxygen tubing with or without No-Crush Lumen.
Our range of Nebulisers with easy to use design are spill proof and provide fast and effective medication delivery. Designed to be used with any standard nebiliser kit and anatomically formed for maximum patient comfort. All products are transparent for easy observation and monitoring. Available as a nebulizer only or with mask and tubing.
The Promed 3-ball, flow-oriented incentive spirometer features three colour-coded balls in chambers to measure inspired volume. The respirometer is used to help improve lung performance in conditions where lung function may be compromised due to various respiratory disorders. This device enables patients to exercise their lungs with the ability to visually examine progress.
Guedel Airway tubes are designed to maintain a secure Oropharyngeal airway for the unconscious patient in emergency pre-hospital care, within the hospital environment and during the anaesthetic peri-operative period.
Made of malleable aluminum covered with plastic sheath, our intubating stylets have a lubricated Satin-Slip surface to help reduce friction between stylet and tracheal tube to allow for easy insertion and withdrawal. They can be easily manipulated to the required shape and remain firm when in use.
Our Yankauer Suction Handles are made of non-toxic k-resin. The tips are glass formed smooth, with side eyes to suit the Yankauer capacity, ensuring maximum suction efficiency and patient comfort. Designed for use with all major suction connecting tubes. Available in crown tip or standard tip, with or without vent, sterile or non-sterile.
We offer a wide range of transport and sterile mucus specimen traps designed to eliminate the possibility of accidental tube separation during specimen transport and prevent contamination from non-sterile suction tubes.