banner-diseases-aquaculture-r3
Shrimp diseases
Infectious hypodermal and hematopoietic necrosis virus (IHHNV)Top
Epidemiology
The IHHNV is a linear single-stranded DNA with an estimated size of 4.1 kb. IHHNV is considered to be a member of the family Parvoviridae, genus Brevidensovirus. Most penaeid species can be infected with IHHNV, including the principal cultured species, P. monodon (black tiger shrimp/prawn), P. vannamei (Pacific white shrimp), and P. stylirostris (Pacific blue shrimp).
A large portion of the IHHNV genome has been found to be inserted in the genome of some genetic lines of P. monodon. There is no evidence that this variant of IHHNV is infectious.
Signs
IHHNV causes the chronic disease runt-deformity syndrome (RDS) in P. vannamei in which reduced, irregular growth and cuticular deformities—rather than mortalities—are the principal effects. IHHNV infection in P. monodon is usually subclinical, but RDS, reduced growth rates, and reduced culture performance have been reported in IHHNV-infected stocks.
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
Infectious myonecrosis virus (IMNV)Top
Epidemiology
IMNV is a totivirus, and closely related to giardia lamblia virus, a member of the family Totiviridae. The genome of IMNV consists of a single, double-stranded RNA molecule of 7560 bp. The principal host species in which IMNV is known to cause significant disease outbreaks and mortalities in farmed populations is P. vannamei.
Signs
Shrimp in the acute phase of IMNV will present focal to extensive white necrotic areas in striated (skeletal) muscles, especially in the distal abdominal segments and tail fan, which can become necrotic and reddened in some shrimp. Severely affected shrimp become moribund, and mortalities can be high immediately following a “stress” event and continue for several days.
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
Taura syndrome virus (TSV)Top
Epidemiology
The genome of TSV consists of a linear, positive-sense single-stranded RNA of 10,205 nucleotides. TSV was listed as an unassigned species in the family Dicistroviridae in the most recent report of the International Committee on Taxonomy of Viruses.
The principal host species in which TSV can cause significant disease outbreaks and mortalities are Penaeus vannamei and P. stylirostris.
Signs
Infected shrimp display anorexia, lethargy, and erratic swimming behavior. They also present opacification of the tail musculature, soft cuticle, and, in naturally occurring infection, a red tail due to the expansion of the red chromatophores. Mortality during this phase can be as high as 95%.
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
Yellow head virus (YHV)Top
Epidemiology
Yellow head virus (YHV) and other genotypes in the yellow head complex are classified by the International Committee on Taxonomy of Viruses as a single species (Gill-associated virus) in the genus Okavirus, family Roniviridae, order Nidovirales. YHV (genotype 1) is one of six known genotypes in the yellow head complex of viruses and is the only known agent of yellow head disease.
YHV is highly infectious for most known species of cultivated penaeid shrimp. Natural infections have been recorded from black tiger shrimp (P. monodon) and P. setiferus.
Signs
The gross signs of shrimps with YHV are:
  • An initial period or exceptionally high feed consumption followed by abrupt cessation of feeding and increased mortality in the population (not always seen).
  • Lethargic or moribund shrimp accumulate at the pond surface and edges with slow to erratic swimming behavior.
  • The region of the cephalothorax may be yellowish due to the light yellow color of the underlying and abnormally soft hepatopancreas (not always seen).
  • Overall body color always of abnormally light or bleached appearance.
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
White spot syndrome virus (WSSV)
Epidemiology
White spot syndrome virus (WSSV) is a double-stranded rod-shaped DNA virus that belongs to the family Nimaviridae, genus Whispovirus. WSSV has an extremely wide host range. The virus can infect a wide range of aquatic crustaceans, especially decapod, including marine, brackish and freshwater prawns, crabs, crayfish, and lobsters.
Signs
The clinical signs of infected penaeid shrimps are:
  • Sudden reduction in feeding, exhibiting lethargic behavior
  • Red discoloration in P. monodon, L. vannamei, and L. stylirostris.
  • Soft, loose shells
  • White spots 0.5 to 2 mm under cuticle (less common in Western Hemisphere penaeids)
  • Up to 100% mortality within 3 days of onset of disease signs.
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
Fish diseases
Infectious salmon anemia virus (ISAV)Top
Epidemiology
ISAV is an enveloped virus, with a genome consisting of eight single-stranded RNA segments with negative polarity. The morphological, physiochemical, and genetic properties of ISAV are consistent with those of the Orthomyxoviridae, and ISAV has been classified as the type species of the genus Isavirus within this virus family.
Natural outbreaks of ISA have only been recorded in farmed Atlantic salmon, and in Coho salmon (Oncorhynchus kisutch) in Chile. In Atlantic salmon, disease outbreaks are mainly reported in seawater cages, and only a few cases have been reported in the freshwater stage, including one case in yolk sac fry.
Signs
The clinical signs of ISAV are:
  • Increased mortality
  • Lethargic fish
  • Gill pallor due to anemia
  • Hemorrhage in the anterior eye chamber
  • Exophthalmia
Regulatory requirements vary by country; products may not be available in your geographic area.
Ask about our solutions ›
For Veterinary Use Only.