Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Support your water and wastewater analysis laboratory needs with Thermo Scientific Orion AquaMate Spectrophotometers. AquaMate Vis and UV-Vis Spectrophotometers combine innovative technology, compact design, and over 260 preprogrammed methods to help attain accurate water analysis.
Depending on the needs of your lab, choose the AquaMate 7100 Vis Spectrophotometer for 325 to 1100 nm wavelengths or the AquaMate 8100 UV-Vis with full 190 to 1100 nm wavelengths for a greater range of measurement options. The AquaMate preprogrammed methods are compatible with many leading reagent manufacturers, including Thermo Scientific Orion AQUAfast reagents.
AquaMate 8100 UV-Vis Spectrophotometer
| Absolute certainty for high-impact confidence
|
AquaMate 7100 Vis Spectrophotometer | Advanced water analysis that meets regulatory methods
|
Chemical species | Method Description | Method | Form Factor | Range | Shop |
Alkalinity | -M, Acid/Indicator Method, 100 tests | AC2002 | Tablet | 5 – 200 mg/L | |
-P, Acid/Indicator Method, | AC3002P | Tablet | 5 – 300 mg/L | ||
Aluminum | Eriochrome Cyanine R Method, 50 tests | AC2027 | Tablet | 0.01 – 0.3 mg/L | |
Eriochrome Cyanine R Method, 100 tests | AC4P27 | Powder & Liquid | 0.01 – 0.25 mg/L | ||
Ammonia as Nitrogen | High Range, Salicylate Method, 50 tubes | ACR011 | Reaction Tube | 1 – 50 mg/L | |
Indophenol Phenate Method, 50 tests | AC2012 | Tablet | 0.02 – 1 mg/L | ||
Low Range, Salicylate Method, 50 tests | ACR012 | Reaction Tube | 0.02 – 2.5 mg/L | ||
Salicylate Method, 100 tests | AC4P12 | Powder | 0.01 – 0.8 mg/L | ||
Bromine | DPD Method, 100 tests | AC2035 | Tablet | 0.05 – 13 mg/L | |
Chloride | Silver Nitrate/Turbidity Method, 50 tests | AC2017 | Tablet | 0.5 – 25 mg/L | |
Chlorine Dioxide | DPD Method, 100 tests | AC2099 | Tablet | 0.2 – 11 mg/L | |
Chlorine | Free & Total, DPD Method, 100 tests | AC2070 | Tablet | 0.01 – 6 mg/L | |
Free, DPD Method, 100 tests | AC2071 | Tablet | 0.01 – 6 mg/L | ||
Free, DPD Method, 100 tests | AC4P71 | Powder | 0.02 – 2 mg/L | ||
Free, DPD bulk powder dispenser with one vial x 250 tests | AQ250F | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast free chlorine DPD bulk powder dispenser* | |
Free, DPD powder refill for AQ250F dispenser, 1 vial x 250 tests | AC71P1 | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast free chlorine DPD powder refill for AQ250F dispenser* | |
Free, DPD powder refill for AQ250F dispenser, 2 vials x 250 tests each | AC71P2 | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast free chlorine DPD powder refill for AQ250F dispenser* | |
Total, DPD Method, 100 tests | AC2072 | Tablet | 0.01 – 6mg /L | ||
Total, DPD Method, 100 tests | AC4P72 | Powder | 0.02 – 2 mg/L | ||
Total, DPD powder dispenser with one vial x 250 tests | AQ250T | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast total chlorine DPD powder dispenser with one vial x 250 count* | |
Total, DPD powder refill for AQ250T dispenser, 1 vial x 250 tests | AC72P1 | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast total chlorine DPD powder refill for AQ250T dispenser* | |
Total, DPD powder refill for AQ250T dispenser, 2 vials x 250 tests each | AC72P2 | Powder | 0.02 – 2 mg/L Cl2 | AQUAfast free chlorine DPD powder refill for AQ250F dispenser* | |
Total, High Range, KI/Acid Method, 100 tests | AC3072 | Tablet | 5 – 200 mg/L | AQUAfast chlorine (total) HR & sodium chlorate reagent tablets | |
COD | Low Range, Dichromate Closed Reflux Method, 25 Tests | CODL00 | Digestion Tube | 0 – 150mg/L | |
Low Range, Dichromate Closed Reflux Method, 150 Tests | CODL150 | Digestion Tube | 0 – 150mg/L | ||
Mid Range, Dichromate Closed Reflux Method, 25 Tests | CODH00 | Digestion Tube | 0 – 1,500mg/L | ||
Mid Range,Dichromate Closed Reflux Method, 150 Tests | CODM150 | Digestion Tube | 0 – 1,500mg/L | ||
High Range, Dichromate Closed Reflux Method, 25 Tests | CODHP0 | Digestion Tube | 0 – 15,000mg/L | ||
High Range, Dichromate Closed Reflux Method, 150 Tests | CODH150 | Digestion Tube | 0 – 15,000mg/L | AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range* | |
Copper | Free & Total, Biquinoline, 50 tests | AC2029 | Tablet | 0.05 - 5mg/L | |
Free, Bicinchoninate, 100 tests | AC4P29 | Powder | 0.05 – 5mg/L | ||
Cyanuric Acid | Melamine Method, 100 tests | AC2098 | Tablet | 0 – 160 mg/L | |
Fluoride | SPADNS Kit Method, 50 tests | AC2009 | Liquid | 0.05 – 2 mg/L | |
Hardness | Total, Method, 100 tests | AC3032T | Tablet | 2 – 50 mg/L | AQUAfast hardness (total) reagent tablets, Metallphthalein Method |
Hydrazine | 4-(Dimethyl-amino)-Benzaldehyde Method, 30 tests | AC2030 | Powder | 0.05 - 0.5 mg/L | |
Iron | (II & III), PPST Method, 100 tests | AC2078 | Tablet | 0.02 – 1 mg/L | |
Ferro, 1,10-Phenanthroline Method, 100 packs | AC4P78 | Powder | 0.02 – 3 mg/L | ||
Total, TPTZ Method, 100 tests | AC4P79 | Powder | 0.02 – 1.8 mg/L | ||
Manganese | Formaldoxime Method, 50 tests | AC2055 | Tablet | 0.2 – 4 mg/L | |
High Range, Periodate Oxidation Method, 100 tests | AC4P55 | Powder | 0.1 – 18 mg/L | ||
Low Range, PAN Method, 100 tests | AC4P54 | Digestion Tubes and Liquid | 0.01 – 0.7mg/L | ||
Molybdate / Molybdenum | Mercaptoacetic Acid Method, 100 tests | AC4P42 | Powder | 0.5 – 66 mg/L | |
Nitrate as Nitrogen (N) | Chromotropic Acid Method, 50 tests | ACR007 | Reaction Tube | 1 – 30 mg/L | |
Zinc Reduction Method, 50 tests | AC2007 | Tablet | 0.08 – 1 mg/L | ||
Diazotization (Azo) Method, 100 tests | AC4P46 | Powder | 0.01 – 0.3 mg/L | ||
Diazotization (NED) Method, 100 tests | AC2046 | Tablet | 0.01 – 0.5 mg/L | ||
Nitrogen | Total, High Range, Persulfate Digestion Method, 50 tests | ACD007 | Digestion Tube | 5 – 150 mg/L | |
Total, Low Range, Persulfate Digestion Method, 50 pack | ACD004 | Digestion Tube | 0.5 – 25 mg/L | ||
pH | pH, Phenol Red Method, 100 tests | AC2001 | Tablet | 6.5 - 8.4 pH | |
pH, Phenol Red Method, 30 tests | AC3001 | Liquid | 6.5 – 8.4 pH | ||
Phosphorous as P | Acid Hydrolyzable, Acid Digestion/Ascorbic Acid Method, 50 pack | ACD095AH | Digestion Tube | 0.02 – 1.6 mg/L | |
Total, Persulfate Digestion/Ascorbic Acid Method, 50 pack | ACD095 | Digestion Tube | 0.02 – 1 mg/L | ||
Phosphate | Phosphate, Ortho, Phosphomolybdenum/Ascorbic Acid Method, 100 tests | AC4P95 | Powder | 0.06 – 2.5 mg/L | |
Ortho, Phosphomolybdenum/Ascorbic Acid Method, 50 tubes | ACR095 | Reaction Tube | 0.06 – 5mg/L | ||
Ortho, Phosphomolybdic Acid/Ascorbic Acid Method, 50 tests | AC2095-WA | Tablet | 0.05 – 4 mg/L | ||
Ortho, Vanadomolybdate/Ascorbid Acid Method, 50 tests | AC2096 | Tablet | 1 – 80 mg/L | ||
Silica | High Range, Silicomolybdate Method, 100 tests | AC4P60 | Powder | 1 – 90 mg/L | |
Phosphate Removal for AC2060 Method, 100 tablets | AC2061 | Tablet | --- | ||
Silicomolybdate, Heteropoly Blue Method, 50 tests | AC2060 | Tablet | 0.05 – 4 mg/L | ||
Sodium Hypochlorite | KI/Acid Method, 100 tests | AC3072 | Tablet | 0.02 – 16% | AQUAfast II Chemistries Test Kits |
Sulfate | Barium Sulfate/Turbidity Method, 100 tests | AC4P82 | Powder | 5 – 100 mg/L | |
Sulfide | DPD/Catalyst Method, 50 tests | AC2016 | Tablet | 0.04 – 0.5 mg/L | |
Zinc | Zincon Method, 50 tests | AC2065 | Tablet | 0.02 – 1 mg/L |
Accessories for AquaMate 7100 & 8100 Spectrophotometers | ||
Chlorine Primary Standard kit, 1.5 mg/L | ![]() |
|
Vial Cleaning Brush, 24 mm vial | ![]() |
|
AQUAfast 24mm Round Vials for Colorimeters and Spectrophotometers |
| |
| ||
Vial, Cuvette and Test Tube Holders for AquaMate AQ7100 and AQ8100 Spectrophotometers |
| |
Replacement Power Cords & Cables for AQUAMate AQ7100 and AQ8100 Spectrophotometers | ![]() |
|
Printer and Supplies for AQ7100 and AQ8100 Spectrophotometers |
| |
Calibration Accessories for AQ7100 and AQ8100 Spectrophotometers |
| |
| ||
Accessories specific to AquaMate 7000 & 8000 Series Spectrophotometers | ||
| ||
Replacement Power Cords & Cables for AquaMate AQ7000 and AQ8000 Spectrophotometers |
| |
| ||
|
Get answers to your top water analysis and water purification questions and see products in action. Learn about:
· pH & Multiparameter Benchtop & Portable Meters
· Electrodes (pH Electrodes, ISE, Conductivity, DO)
· Potentiometric Titrators
· Spectrophotometers
· Water Purification Systems