Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The evolution of rapid culture media-based methods has been made possible by the development of enhanced performance enrichment broths and the use of chromogens in agar media.
All Thermo Scientific™ Precis™ Detection Methods follow the principal of a single overnight enrichment, a single chromogenic agar plate and a single, simple confirmation step. Compared to traditional culture-based methods, Precis Methods are simpler to run and generate results in a reduced timeframe enabling quicker actions and shorter product hold times.
The Thermo Scientific Listeria Precis Enumeration Method and Thermo Scientific™ Brilliance™ chromogenic agar enumeration methods are equally simple. They replace standard methods, that require multiple dilution and plating steps, with a single dilution and single plating step, followed by simple confirmation.
To find out more about our complete range of solutions for the detection of Salmonella species in foods and related sample types download our free guide.