Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Our selection of field and bench calibrators sources, simulates, and measures pressure, temperature, and electrical signals with exceptional precision.
Whether you're calibrating instruments, troubleshooting a problem, or running routine maintenance, Fluke’s range of multifunction calibrators can help you get the job done faster. They are capable of accomplishing so many different tasks and do them quickly and efficiently; it's the only process calibrator you’ll need to carry.
The Fluke 753 and 754 documenting process calibrators offer powerful multifunction documenting calibration that you can use to download procedures, lists, and instructions created with software or upload data for printing, archiving, and analysis. The powerful, built-in HART® interface in the 754 is capable of performing nearly all the day-to-day tasks you now perform with a separate communicator.
These documenting calibrators do the work of several tools. It sources, simulates, and measures pressure, temperature, and electrical signals with one rugged, hand-held device. For documentation, they automate calibration procedures and captures your data. And, of course, it helps you meet rigorous standards like ISO 9000, FDA, EPA, and OSHA regulations. Plus, the new improved graphical screen, Li-Ion battery for longer life, USB port, and accessories help you work smarter and faster.
Documenting process calibration with HART® | Documenting process calibration | Pressure modules and accessories |
---|---|---|
![]() | ![]() | ![]() |
Rugged, reliable tool for calibrating, maintaining and troubleshooting HART® and other instrumentation. | Rugged handheld tool for sourcing, simulating and measuring pressure, temperature and electrical signals. | A full range of gauge, differential and absolute pressure modules, pumps hoses and fittings. |
Fluke 754 Documenting Process Calibrator with HART® | Fluke 753 Documenting Process Calibrator | Fluke 750P Pressure Modules |
Designed specifically for the process industry with broad workload coverage, calibration power, and unsurpassed accuracy, Fluke’s range of multifunction process calibration tools has you covered.
Precision multifunction process calibrator | Multifunction process calibrator | IS multifunction process calibrator |
---|---|---|
![]() | ![]() | ![]() |
This has all the features of the Fluke 725 plus enhanced accuracy, pulse count sourcing, and pulse measurement, totalizing pressure switch test and % error calculation in one compact package. | Here is a powerful, easy-to-use field calibrator to test and calibrate almost any parameter. Measure, source, and simulate volts, mA, thermocouples, RTDs, frequency, ohms, and pressure to calibrate transmitters. | ur easy-to-use, intrinsically safe field calibrator can calibrate almost any process instrument needing service where explosive gases might be present and includes all the features of the 725. |
Fluke 726 Precision Multifunction Process Calibrator | Fluke 725 Multifunction Process Calibrator | Fluke 725Ex IS Multifunction Process Calibrator |
The Fluke 753 and 754 documenting process calibrators offer powerful multifunction documenting calibration that you can use to download procedures, lists, and instructions created with software or upload data for printing, archiving, and analysis. The powerful, built-in HART® interface in the 754 is capable of performing nearly all the day-to-day tasks you now perform with a separate communicator.
These documenting calibrators do the work of several tools. It sources, simulates, and measures pressure, temperature, and electrical signals with one rugged, hand-held device. For documentation, they automate calibration procedures and captures your data. And, of course, it helps you meet rigorous standards like ISO 9000, FDA, EPA, and OSHA regulations. Plus, the new improved graphical screen, Li-Ion battery for longer life, USB port, and accessories help you work smarter and faster.
Documenting process calibration with HART® | Documenting process calibration | Pressure modules and accessories |
---|---|---|
![]() | ![]() | ![]() |
Rugged, reliable tool for calibrating, maintaining and troubleshooting HART® and other instrumentation. | Rugged handheld tool for sourcing, simulating and measuring pressure, temperature and electrical signals. | A full range of gauge, differential and absolute pressure modules, pumps hoses and fittings. |
Fluke 754 Documenting Process Calibrator with HART® | Fluke 753 Documenting Process Calibrator | Fluke 750P Pressure Modules |
Designed specifically for the process industry with broad workload coverage, calibration power, and unsurpassed accuracy, Fluke’s range of multifunction process calibration tools has you covered.
Precision multifunction process calibrator | Multifunction process calibrator | IS multifunction process calibrator |
---|---|---|
![]() | ![]() | ![]() |
This has all the features of the Fluke 725 plus enhanced accuracy, pulse count sourcing, and pulse measurement, totalizing pressure switch test and % error calculation in one compact package. | Here is a powerful, easy-to-use field calibrator to test and calibrate almost any parameter. Measure, source, and simulate volts, mA, thermocouples, RTDs, frequency, ohms, and pressure to calibrate transmitters. | ur easy-to-use, intrinsically safe field calibrator can calibrate almost any process instrument needing service where explosive gases might be present and includes all the features of the 725. |
Fluke 726 Precision Multifunction Process Calibrator | Fluke 725 Multifunction Process Calibrator | Fluke 725Ex IS Multifunction Process Calibrator |