Browse solutions for playback/simulation
Making sounds and vibrations perceptible
From the pure, aurally accurate playback of binaural recordings to the realistic reconstruction of sound fields and background noise – HEAD Acoustic's playback and simulation systems make sounds and vibrations perceptible.
Aurally accurate playback
labP2
labP2 is a binaural headphone equalizer (equalization: FF, ID, DF, USER) that enables aurally accurate playback of binaural recordings via one or two separately equalized headphones. It can be used in USB mode with a PC or as part of a HEADlab. In addition to the mobile version, the labP2 is available as the labP2-V1 for desktop and rack applications.
labP2 can play back binaural recordings with the correct equalization. labP2 features two discrete headphone outputs that can be calibrated and equalized independently of each other. The sound playback quality with separately equalized headphones is comparable to the original sound field.
labO2
labO2 is a 2-channel playback equalizer with line outputs (equalizer: FF, ID, DF, USER). It allows aurally accurate playback of binaural recordings when using the loaded filters to incorporate the human auditory impression in the assessment of sounds.
In addition to the mobile version labO2, a rack version labO2-V1 is available. This also has an integrated headphone connection.
The labO2 equalizer can be used in USB mode or as a playback module in a HEADlab system. It allows for the connection of HDA IV headphone amplifiers and power amplifiers for playback via speakers, subwoofers, shakers, etc. Using several labP2 and labO2, it is possible to form a larger synchronized playback system.
labO2-V1
labO2-V1 is a universally usable 2-channel equalizer for playback via headphones, speakers, subwoofers, or shakers. The 2-channel equalizer labO2-V1 has an output for electrodynamic headphones and a line output (equalizer: FF, ID, DF, USER). Therefore, labO2-V1 combines the best of both worlds: The integrated headphone equalizer of labP2 and the excitation function of labO2.
The equalizer function allows binaural recordings to be played back with the correct equalization when using the loaded filters. This aurally accurate playback allows the human auditory impression to be incorporated in the assessment of sounds. labO2-V1 is a universal rack version.
HD Headphones
To reproduce binaural recordings accurately and realistically, you need high-quality headphones perfectly matched to and equalized by the playback devices you use. HEAD acoustics offers a sensible selection of open and closed electro-dynamic headphones in three price and quality levels for all conceivable demands and requirements. The headphones have ideal sound characteristics, a natural sonic profile, and high timbre faithfulness. 
Depending on the headphone model and the front end, HEAD acoustics offers an individual level calibration and headphone equalization or a model-specific standard equalization to create a sound impression comparable to the original sound field during playback.
HPL
Realistic playback is essential for adequately evaluating product sound quality in a listening studio.
Our HPL active high precision speaker cabinet enables recorded sound to be played back with incredible accuracy. The speakers come very close to the perfect sound transducer and are ideal for equalized playback in listening studios. The results of hearing tests can thus be optimally evaluated. The realistic sound playback allows various scenarios to be run through interactively and any variations to be discussed with management.
The design is much sturdier than conventional wooden housings and virtually free of resonance and vibration. The front and rear panels are made from robust aluminum. The base contains additional spring damping.
HSW
The HSW I and HSW II.1 subwoofers enhance high-quality playback via headphones or speakers. They allow low-frequency components of airborne sound to be taken into account in perceptive sound assessment. This means that perception of the sound played back matches up more closely with reality. To ensure aurally accurate playback, the subwoofers are filtered with a labO2 / labO2-V1 system.
HSW I
In a listening studio, the HSW I subwoofer set completes playback through headphones or speakers with efficiency optimized for the lower frequency range.
HSW II.1
The HSW II.1 subwoofer is ideal for sound playback in a vehicle setting (SoundCar, for example). Its relatively small size means it can be installed in the trunk of the vehicle.
HDA IV
HDA IV is a high-quality distribution amplifier (HDA stands for "Headphone Distribution Amplifier") to which multiple headphones can be connected for binaural playback. Using HDA IV with labO2-V1 playback equalizer ensures acoustically accurate playback. With HDA IV distribution amplifier, custom level calibration for each individual headphone is a standard feature. HDA IV is particularly suited to jury testing with SQala software.
Two versions of the distribution amplifier are available:
  • HDA IV.1: Up to 4 dynamic headphones can be connected
  • HDA IV.2: Up to 8 dynamic headphones can be connected
BPU Bundle
With the BPU Bundle, you can easily take advantage of the benefits of aurally accurate playback - whether alone at a PC or in a listening studio. The playback unit and one of the included headphones, HD CL I.1 (closed) and HD OP I.1 (open), are already matched to each other at the factory by the model-specific standard equalization filter. The playback thus conveys a listening impression comparable to the original sound field without any further configuration.
The headphones HD CL I.1 (closed) and HD OP I.1 (open) serve most playback scenarios and impress with their balanced and contoured sound.
Background noise simulation
Background noise impairs speech intelligibility during phone conversations or the quality of voice recognition systems. In order to evaluate these devices qualitatively, they should be tested with realistic and reproducible background noise. Corresponding systems for background noise simulation consist of hardware components for recording, equalization, and playback as well as a main control software covering the functionalities.
3PASS lab
Fully-automated background noise simulation for testing smartphones and other communication devices.
The 3PASS lab exactly reproduces background noises in different test rooms in terms of level, frequency as well as spatial and temporal distribution in the area of the device under test. Application-specific microphone arrays (MSA I or MSA II) allow to record sound fields and to equalize the multi-channel background noise simulation system. Here, the equalization of the whole system is digital and fully-automated – one of the main advantages of 3PASS lab.
3PASS flex
Fully-automated background noise simulation to test systems with complex microphone setups.
The fully-automated software 3PASS flex enables to simulate precisely background noise in different test rooms or in vehicle cabins in terms of level, frequency as well as spatial and temporal distribution in the area of the device under test. Application-specific microphone arrays (MSA I or MSA II) allow to record sound fields and to equalize the multi-channel background noise simulation system. The benefit of 3PASS flex: Equalization of the whole system is conducted fully-automated and digital.
The playback and recording system is particularly suited for testing systems with multiple distributed microphones or microphone arrays under real-world conditions. These include applications like car hands-free systems, In-Car Communication (ICC) systems, voice-controlled smart home devices, smart speakers, conference systems and televisions. When testing these systems, a good sound field reproduction must be achieved simultaneously at the positions of all microphones used. Therefore, the multi-point noise simulation (MPNS) method is applied.
3PASS reverb
Simulate room reverberation realistically and reproducibly.
The software option 3PASS reverb extends these two software solutions with the ability to realistically and reproducibly simulate room reverberation of a single talker at the position of the device under test in a laboratory.
While an artificial head plays back the direct sound signal, the background noise system covers playback of the reverberation components based on the impulse response from the original room. 3PASS reverb operates with available equalized setups from 3PASS lab and 3PASS flex.
The software option 3PAS reverb in combination with the simulation software 3PASS lab or 3PASS flex, one of the microphone arrays MSA I or MSA II (recording, equalization), the hardware interface labBGN (equalization, playback) as well as loudspeakers and power amplifiers form a powerful complete system for the simulation of background noise and reverberation.
HAE-BGN
Background noise playback and semi-automated equalization in a lab.
The software HAE-BGN supports the semi-automated equalization of 4.1 loudspeaker arrangements in a lab. Additionally, it enables the automated playback of background noise and thus a realistic simulation. Based on the equalization, it is possible to reproduce previously recorded noises in level and spectrum at an arbitrary location in the vehicle (e.g. DUT microphone). This allows precise reproduction of the sound field at this location.
HAE-car
Background noise playback and semi-automated equalization in the vehicle.
The software HAE-car allows the semi-automated equalization and enables automated playback of background noises in the vehicle cabin. Based on the equalization, it is possible to reproduce previously recorded noises in level and spectrum at an arbitrary location in the vehicle (e.g. DUT microphone). This allows precise reproduction of the sound field at this location.
labBGN
Hardware platform for simulation of true-to-reality background noise scenarios.
labBGN is the key hardware component for operating HEAD acoustics background noise simulation systems. With the hardware platform and the corresponding simulation software (3PASS lab, 3PASS flex, 3PASS reverb, HAE-BGN, HAE-car), it is possible to record, equalize and realistically playback background noises as well as reverberation (impulse responses) – reproducible across different laboratories.
labBGN provides a maximum of ten channels: Thus, users are able to connect e.g. eight loudspeakers and one stereo subwoofer to the labBGN concurrently.
MSA I & MSA II
Microphone arrays for three-dimensional noise recordings.
The microphone arrays MSA I (asymmetrical) or MSA II (symmetrical) are essential system components for operating the background noise simulation systems from HEAD acoustics. Controlled by the corresponding simulation software (3PASS lab, 3PASS flex, 3PASS reverb) both microphone arrays enable to record, equalize and realistically reproduce background noise as well as reverberation (impulse responses) – reproducibly across different laboratories or vehicle cabins.
Within this system setup, a microphone array is necessary for the automated digital equalization of the whole system. In addition to equalization, the microphone arrays allow users to record three-dimensional noise scenarios that are required for realistic tests of communication devices.
Discuss your needs with an expert
Speak to our technical experts to discuss your application requirements