Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The HEAD VISOR measurement system comprises the software and a microphone array. The acoustic camera uses beamforming technology to pinpoint sound sources in real time. Various microphone arrays are available for different applications. Some are scalable and customisable and all can be activated and controlled with HEAD VISOR software.
As soon as the microphone array is switched on, HEAD VISOR software provides video images with simultaneously overlaid, high-quality source mapping. This feature allows you to obtain the necessary information about the sound source without any time delay.
All three hardware versions combine beamforming with MultipleEye technology, using three cameras to record the video images in real time. This process enables distances to all points in the image to be determined with exceptional precision and allows continuous, synchronous video images in real time.
HEAD VISOR VMA II.1 has 56 microphones positioned in a spiral pattern across a diameter of 80 cm and is easy to transport. The spiral arrangement allows distances of 30 cm to 200 m between the microphone array and sound source, with dynamics of 13 dB (standard beamforming). Advanced algorithms extend the dynamics to 20–30 dB in a frequency range of 300 Hz–20 kHz.
The HEAD VISOR VMA III.0 array, with 112 microphones and a diameter of 3 m, is suitable for installation on a wall or ceiling, which means it can be used in e.g. wind tunnels. The 14 arms allow dynamics of 10 dB (standard beamforming) or up to 30 dB (advanced algorithms) in a frequency range of 200 Hz–20 kHz.
HEAD VISOR flex is a modular microphone array for sound source location. Its configuration, size, layout and number of microphones can be adjusted to suit your needs. HEAD VISOR flex delivers extremely high accuracy for localising sounds over large distances or at low frequencies of less than 100 Hz.
The near-field HEAD VISOR Probe helps with investigating stationary sounds at frequencies below 600 Hz and acts as a supplement to the microphone arrays. Sound is detected directionally via two microphones at the tip of the probe. MultipleEye technology determines the current position of the probe in real time. The probe therefore allows specific sub-areas to be calibrated. The near-field HEAD VISOR Probe can be connected to the array, which allows a frequency range of 20 Hz–20 kHz to be evaluated.
HEAD VISOR is the software application for localising sound sources online in real time. It combines functions such as recording, playback, analysis and reporting in one package. Coupling it with the various microphone arrays forms an acoustic camera, a beamforming system, which allows detailed insight into the acoustics of complex measurement subjects.
The software is ready to use as soon as the microphone array has been switched on, with recording starting automatically. Thanks to the permanently active FreezeBuffer, users can also easily analyse an acoustic event at a later point in time.
HEAD VISOR is a powerful and easy-to-use tool, with all the key applications available for the user to view at all times. This allows tracking the effects of modifications, filters and changes to sound sources in real time.
HEAD VISOR remote, the integrated remote control, offers a number of additional functions. Multiple microphone arrays, HEADlab systems, computer or memory systems can be centrally controlled, managed and synchronised. Automated measurements and analyses can be conducted while ArtemiS SUITE components can be integrated in the evaluation.
Speak to our technical experts to discuss your application requirements