Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Smart Dilutor W is the solution for labs seeking full processing capacity and traceability in gravimetric dilutions or liquid dispensing. This device ensures precision, rapidity, sample handling sterility, and detailed tracking of procedures.
The IUL Smart Dilutor weighs samples and dispenses an accurate amount of broth, diluting the sample to a configurable dilution factor. This makes sample preparation easier and more efficient, getting them ready to be homogenized for further testing.
The system’s highlights include:
Catalog number | Description | |
---|---|---|
IUL7900 | Smart Dilutor W with one pump | Request quote › |
IUL7901 | Smart Dilutor W with two pumps | Request quote › |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.