The LanthaScreen™ kinase activity assay toolbox has been designed with four key goals in mind:
  • To provide users the ability to rapidly apply TR-FRET technology 
  • To develop a toolbox of reagents to support flexible assay configuration
  • To provide a TR-FRET technology adapted to multiple instrument platforms
  • To provide custom assays "on demand"
View kinase specific protocols, compatibility, and assay component selection table
Our LanthaScreen kinase activity assay toolbox of 'mix and match' assay reagents is designed to enable rapid assay configuration. Our portfolio of phospho-specific antibodies, matched fluorescent peptide substrates and in-house produced kinases have been tested in >200 different kinase assays to date.
Request Assay Information ›        Use Assay Maker™ ›
LanthaScreen Activity Assay Mechanism
Schematic Illustration of a LanthaScreen Kinase Activity Assay
LanthaScreen Assay Schematic
In a LanthaScreen kinase activity assay, kinase, fluorescein-labeled substrate, and ATP are allowed to react. Then EDTA (to stop the reaction) and terbium-labeled antibody (to detect phosphorylated product) are added. In a LanthaScreen kinase reaction, the antibody associates with the phosphorylated fluorescein labeled substrate resulting in an increased TR-FRET value. The TR-FRET value is a dimensionless number that is calculated as the ratio of the acceptor (fluorescein) signal to the donor (terbium) signal. The amount of antibody that is bound to the tracer is directly proportional to the amount of phosphorylated substrate present, and in this manner, kinase activity can be detected and measured by an increase in the TR-FRET value.
Schematic of a LanthaScreen activity assay using secondary antibodies
LanthaScreen Secondary Antibody Application
 In a kinase assay utilizing the terbium labeled anti-Mouse antibody generic reagent, the assay is very similar to the direct format with the exception that the phospho-specific antibody is not labeled and the detection step is performed by the addition of the terbium labeled anti-Mouse antibody. This approach can be applied to target classes beyond kinases.
For Research Use Only. Not for use in diagnostic procedures.