Gibco CTS DynaCellect Magnetic Separation System
Consumables and software
The Gibco™ CTS™ DynaCellect™ Cell Isolation Kit and the Gibco CTS DynaCellect Bead Removal Kit are specifically designed, single-use only consumables for a range of cell processing applications such as isolation, depletion, and bead removal. Each kit has input/output sterile weldable tubes for flexible setup configurations.
Contact us    View 3D tour
The CTS DynaCellect Magnetic Separation System is able to scale from small to large volumes for process development and clinical and commercial manufacturing.
CTS DynaCellect Cell Isolation Kit 
The CTS DynaCellect Cell Isolation Kit is specifically designed for the isolation and activation of immune cells in cell processing applications.
Specifications
Attribute
Description
Maximum capacity
1,000 mL
8 weldable lines for input and output bags
D-E-H-P-free PVC (150 mm length, 2.4 mm ID, 4.0 mm OD)
Sterile air filter
0.20 micron
Sterilization method
Gamma irradiation
Note: Instrument and Consumable Regulatory Support File available upon request at thermofisher.com/regulatory.
CTS DynaCellect Bead Removal Kit
The Gibco CTS DynaCellect Bead Removal Kit is specifically designed for bead removal from immune cells for cell processing applications.
Specifications
Attribute
Description
Maximum capacity
Continuous processing
4 weldable lines for input and output bags
D-E-H-P-free PVC (150 mm length, 2.4 mm ID, 4.0 mm OD)
Sterile air filter
0.20 micron
Sterilization method
Gamma irradiation
Note: Instrument and Consumable Regulatory Support File available upon request at thermofisher.com/regulatory.
Download the product specification sheet ›
Software
The CTS DynaCellect Magnetic Separation System comes with two protocols: isolation and bead removal. If you want to customize a protocol, the intuitive, user-programmable interface enables you to create and manage all instrument actions. When customizing the protocol, you can: 
  • Engage or disengage the magnet
  • Open or close the pinched valves
  • Change the pump speed from ~2–150 mL/min
  • Change the specified volume of the pump
  • Change the rocker angle from 1–30°
  • Change the rocker speed from 1–30 RPM
  • Change the air pressure sensor (max 2 bar)
Once you have an optimized protocol, you can switch to clinical manufacturing mode, which will lock down your optimized protocol. In this mode you can perform the following standardized cell processing steps:
  • Cell isolation
  • Cell depletion
  • Bead:cell binding and incubation
  • Bead removal
  • Ready for sterile sampling for IPC/QC
  • Operator mode (locked software)
Clinical manufacturing mode also includes an optional software module that supports: 
  • Enablement of 21 CFR part 11 compliance (software)
  • DeltaV communication via OPC UA
  • Providing a summary file (sample ID, run ID, SUT lot, bead lot, operator, date)
  • Preparation of barcoding through the USB port
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals