Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Stay at the leading edge of imaging and documentation systems for a wide range of highly demanding applications, including chemiluminescence and spectral fluorescence, through state-of-the-art camera technology and optical innovations. Sensitivity, resolution, and robustness are at the core of Uvitech product developments, designed to provide industry specialists with data they can trust and rely on.
Essential | Uvidoc | Firereader | Platinum | |
---|---|---|---|---|
Native resolution | 3 megapixels | 6 megapixels | 6 megapixels | 6 megapixels |
Extended resolution | 6 megapixels | 10 megapixels | 10 megapixels | 10 megapixels |
Dynamic range | 4.0 OD, 16-bit CCD | 4.0 OD, 16-bit CCD | 4.0 OD, 16-bit CCD | 4.0 OD, 16-bit CCD |
Filter wheel | 1 | 3 | 6 | 7 |
Transilluminator | ||||
Epi-white light | ||||
Epi-blue light | - | - | - | |
Epi-UV 254/365 nm | - | - | Optional | - |
Epi-fluorescence | - | - | - | Optional |
Upgradreable | - | - | - | Chemiluminescence |
Datasheet › | Datasheet › | Datasheet › | Datasheet › |
Essential V6
The Essential V6 is the most compact yet high-end imaging platform for DNA, RNA, and protein gel imaging, boasting a 3-megapixel, 16-bit sensor offering massive 65,535 gray scale ideal for lab imaging routine, research-level data collection, and accurate protein quantitative analysis.
Uvidoc HD6
The Uvidoc HD6 is a unique, stand-alone image-acquisition platform featuring a real on-board computer and giant 12.2″ HD touch screen, boasting a patented 1-touch concept for unrivalled speed, simplicity, and user-friendliness. This is ideal for crowded labs looking for automation, ease-of-use, and high sensitivity.
FireReader V10
The FireReader V10 is a super powerful, multi-application, full imaging platform offering up to 7 different illumination sources and countless application possibilities. Boasting a huge 6-megapixel sensor, the FireReader V10 delivers up to 65,535 gray levels, ideal for publication-level images and research-level data.
Platinum Q9
The Platinum Q9 is a Chemiluminescence-upgradeable geldoc with the Alliance Q9 Q-smart darkroom. It comes with a choice of over 6 interchangeable plug-n-play transilluminators and 17 emission filters, a massive 6-megapixel resolution, and a 16-bit scientific sensor delivering up to 65,535 shades of gray.
Q9 Advanced | Q9 Manual | |
---|---|---|
Optics | Motorised | Manual (pre-calibrated) |
Native resolution | 9.2 megapixels | |
Extended resolution | 18 megapixels | |
Dynamic range | >4.8 OD, 16-bit CCD (65,536 shades of gray) | |
Filter wheel | 7-position, motorised | |
Camera cooling | –60°C regulated, 3-stage peltier | |
Lens | f/0.80 panoramic | |
Field of view | 24 x 20 cm | |
Epi-white | ||
Transilluminator | Optional, large choice | |
Spectral fluorescence | Optional, large choice (8 excitation, 18 emission) | |
Epi-UV 365nm | Optional |
Alliance Q9 Advanced–Manual
This system offers a low-cost option with the power of the Q9-series. Q9 features the same camera and darkroom specifications, plug-n-play upgradeability, and femtogram-level sensitivity as the Q9 Advanced, yet with pre-calibrated optics, allowing to take full manual control of the imager’s exceptional potential.
Alliance Q9 Advanced–Automatic
This system features automation, power, and versatility. Expect the best of what the future of imaging has to offer: highest resolution and sensitivity on the market, up to 17 illumination sources including 8 spectral fluorescence wavelengths, 17 filters to choose from, and full plug-n-play upgradeability at all times.
Alliance Q9 Touch
The all-in-one Alliance features a real on-board computer. Enjoy all Alliance Q9 has to offer in a powerful, remarkably user-friendly, stand-alone system, ideal for high-demanding applications, research, and publications, as well as crowded labs where automation, ease-of-use, and top-sensitivity are required.
For Research Use Only. Not for use in diagnostic procedures.