Cold Storage Solutions
From 5°C to -196°C
Explore our cold storage solutions
Designed with your science in mind
Most laboratories have a very large collection of biological samples from clinical and field studies, some irreplaceable, all representing enormous scientific and financial value for the researcher and the organisation. Hence a key aspect of research revolves around the preservation of samples for examination and analysis at a future date. It is imperative to have a process of storage (short and long term) that is efficient and preserves sample integrity over time.
How can Thermo Fisher Scientific partner with you?
For over 80 years, we have been a trusted and reliable partner in the development of sustainable, high-quality products for cold storage applications ranging from 4 °C high-performance laboratory refrigerators to –196 °C cryogenic freezers. We are committed to designing products and service solutions with your science in mind, helping you be confident on paper and in practice.
Discover why researchers worldwide protect more than 2 billion samples with Thermo Scientific cold storage equipment and consumables. Designed to support your science – across all temperatures.
Our cool tools for your science
Sample collection plasticware
Barcoded tubes
Thermo Scientific™ Matrix™ 2D Barcoded Screwtop Storage Tubes are ideal for applications at temperatures from -80°C to -196°C.
  • Permanent 2D barcodes enable safe, secure tracking of samples
  • Certified SAL of 10-3
  • Certified DNase-, RNase-, pyrogen-, and toxin-free
  • Volumes from 200 μL to 5 mL
  • 7 cap colours available for 0.5 mL and 1 mL sizes
Storage plates
Thermo Scientific™ Nunc™ Storage Plates are polypropylene plates with 48, 96 or 384 wells.
  • Made with high-quality virgin polypropylene resin
  • Certified DNase, RNase and human
  • DNA free
  • ANSI format for compatibility with automated systems
  • Stable at temperatures -80°C to +121°C
  • Sealing options, incl adhesive or heat seals, are available
Cryogenic tubes
Store samples with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic tubes.
  • From general cold storage to the vapour phase of liquid nitrogen
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free cryogenic tubes
  • Available with either internal or external threads.
  • Certified SAL of 10-6
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Sealing equipment
Capper/decapper
Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper helps maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily.
  • Ideal for integration into automated systems
  • Cap and decap an entire rack at a time in approximately 10 seconds
  • Suitable for 48-, and 96- format Thermo Scientific™ Matrix™ and Nunc™ Screw Top tubes
Handheld capper/decapper
The 8-channel Thermo Scientific™ Handheld Capper and Decapper is a practical addition to benchtop processing of screw-top storage tubes.
  • Processes a full rack of Thermo Scientific™ Matrix™ or Nunc™ tubes in less than one minute
  • Caps all tubes to the optimal torque level specified for the tube
  • Lightweight, easy-to-use and ergonomic design
Plate sealers
Thermo Scientific™ ALPS5000™ plate sealing equipment is the next generation of automated plate sealing.
  • Seals up to 2 plates per minute in handfed, benchtop, or high-throughput robotic applications
  • Touchscreen control and parameter customisation
  • Electrically driven with tool-free loading
  • Energy-efficient and energy-conscious
  • Improved reliability and noise reduction
Tracking equipment
High speed barcode reader
Thermo Scientific™ VisionMate™ HSX High Speed barcode reader reads 2D barcoded tubes in as little as 1 second.
  • Can read both 2D and linear rack barcodes (Pre-loaded rack configurations for Matrix™ and Nunc™ racks)
  • Graphical user interface provides multiple options for data viewing
  • IP66 rating provides a critical environment barrier that enables clean room use
Storage equipment
Ultra-low temperature freezers
Thermo Scientific™ Ultra-Low Temperature freezers deliver uncompromising reliability and dependability.
  • Ergonomic design; optimised storage space
  • Energy efficient and environmentally sustainable
  • Vapour compression enables faster Door Opening Recovery speed
  • Meets ISO Class 5.5 
Laboratory fridges
Thermo Scientific™ scientific and medical refrigerators provide reliable, secure storage for all of your samples.
  • Ergonomic features, intuitive controls, whisper-quiet operation
  • Innovative compressor technology
  • Engineered for environmental sustainability 
Laboratory freezers
Thermo Scientific™ freezers are purpose-built to support the demanding requirements of the lab and healthcare facilities.
  • Intuitive controls, user friendly
  • Up to 10x quieter
  • Fast door opening recovery
  • Quality construction
  • ENERGY STAR certified
Cryopreservation systems
Thermo Scientific™ cryopreservation systems comprise a full range of LN2 equipment to enhance long-term sample viability.
  • Advanced insulation features and design innovations ensure exceptional temperature stability
  • Wide range of storage capacities
Five reasons for choosing greener cold storage lab equipment
Discover the benefits of going green.
Monitoring equipment
Remote monitoring
Thermo Scientific™ Smart-Vue™ Pro Remote Monitoring Solution is the next-generation system that is connected by a secure, long range, dedicated network.
  • Sample protection – instant alerts for out-of-range sample conditions
  • Compliance – automatic and continuous data logging
  • Asset/ cost optimisation – can serve as part of your asset management strategy
Wireless tracking
Thermo Scientific™ Smart-Tracker™ wireless datalogging module makes temperature monitoring in cold chain easy.
  • Monitor storage facilities, freezers, refrigerators, heating equipment and packages in transit
  • Bluetooth wireless connection for use in different types of containers and enclosures
Explore our full spectrum of cold storage solutions in 3D
The right equipment for every temperature
We support you with not only world-class products, but also “value beyond product” with our full service offerings and technical support. Click through our curated areas to see how we can support you.
  • Academic
  • Biobanking
  • Cell & Gene Therapy
  • Blood bank
  • Vaccine
  • Molecular diagnostic
Frequently asked questions
Want more information on specific products or workflows?
Feel free to contact us if you have any questions, want more information on specific products and workflows, or wish to discuss your needs. We are always there to partner with you and your lab.