Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Konica Minolta Sensing precise light and display measurement range offers an array of styles and sizes for measuring light accurately. For those on the go, many of these light meters are easily portable and simple to read and understand.
We offer Konica Minolta light measuring equipment that will fit your individual budget and needs, whether you need to measure illuminance or luminance. Light meters are not all the same, as needs differ from application to application, so choose the style and light meter most suited to your individual needs by browsing our options.
Gloss meters are used to measure specular reflection. Specular reflection has a direct relationship between the ratios of incident light and reflected light, and has been used as the basis for standardizing and measuring gloss values. There are numerous International Standards that define the methodology and technical specifications of gloss meters for measurement on different types of materials. Konica Minolta’s gloss meters keep in line with these standards, and are crafted with precision and accuracy in mind.
Konica Minolta light meters can be used in labs to measure chemically reactive substances, the illumination of LEDs and CRTs, for quality control and government testing, and for photography and cinematography. These light and display meters are easy to read and understand, and are designed to fit your budget and needs, whether you need to measure illuminance or luminance.