Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Fume hoods protect laboratory staff from noxious fumes released by acids, dangerous gases, and organic solutions. Harmful and unpleasant chemical fumes are removed from the controlled environment to facilitate a safe and pleasant work environment. Ducted fume hoods channel chemical vapours out of the building using an external fan installed on the roof or on an external wall. TopAir fume hoods can be customised to any size to suit your needs.
TopAir active polypropylene fume hoods are a high-quality system enhanced with innovative technology. The fume hood features a sensor which detects staff standing next to the unit, and accordingly opens and closes the sash automatically.
Variable airflow technology allows the unit’s fan speed to adjust according to the sash position, reducing energy consumption and noise. Upon a failure, the whole interior of the cabinet lights up with a red light, so that staff—including people with hearing issues—can easily detect the failure from a distance.
TopAir polypropylene fume hoods are made of high-quality non-corrosive polypropylene with excellent chemical resistance. They protect laboratory staff from noxious fumes released by acids, dangerous gases, and organic solutions which regular metal hoods may not withstand. Polypropylene increases the product’s tensile strength and improves its thermal characteristics.
TopAir polypropylene fume hoods with wet scrubber feature a quality scrubbing media which collects mist and chemicals and channels them down to the water tank. Its upper eliminator prevents mist from reaching the exhaust fan. The fume hood filters out acids to prevent them from being released into the environment. The scrubber is built-in as an integrated part of the hood.
TopAir’s walk-in fume hoods offer a large front opening designed to house large, heavy, or tall equipment.
TopAir metal fume hoods protect laboratory staff from noxious fumes when working with acids, dangerous gases, and organic solutions. The hood structure is made of epoxy-covered metal, while the internal structure is made of 6 mm high-pressure laminate (HPL).
TopAir educational fume hoods have transparent sides and back for 360° visibility, allowing delivery of educational sessions.
Brochure
Specification sheets