Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a comprehensive selection of TopAir laboratory chemical fume hoods designed to outperform current safety requirements with superior advanced and unique safety features while providing energy and cost efficiency. TopAir fume hoods are used in laboratories worldwide and are valued for their safety, performance, durability, and styling.
Browse our selection of enclosures below including fume hoods, laminar flow clean benches, PCR enclosures, and balance enclosures designed to protect either the operator, process, or both from toxic vapours, fumes, gases, and particulates.
TopAir Systems is a manufacturer of innovative clean air and containment solutions. TopAir clean air solutions stand out for their advanced safety features, and are used in laboratories and manufacturing facilities within chemical/biological plants, universities, research and development facilities, and hospitals, as well as in the electronics, semiconductor, and pharmaceutical industries.
Fume Hoods protect laboratory staff from noxious fumes released by acids, dangerous gas, and organic solutions. Harmful and unpleasant chemical fumes are removed from the controlled environment to facilitate a safe and pleasant work environment using an external fan installed on the roof or on an external wall.
Ductless fume hoods offer flexibility and mobility without sacrificing on safety and quality. TopAir Polypropylene Ductless Fume Hoods provides a safe work environment for lab staff working with certain acids and chemicals without the need for complicated fixed installation.
Horizontal and Vertical Laminar flow clean benches are used to provide product or sample protection for applications that do not require personal or environmental protection. These clean benches provide product protection through the use of controlled laminar airflow and filtration at minimal operational costs.
Balance enclosures are the solution to weighing of potent compounds and chemicals. TopAir Weighing Stations enable precise weighing data up to 4 decimal places.
PCR-UV/HEPA cabinets offer a quality filtering system which provides complete protection for your samples from contamination. PCR cabinets are used for genomics, proteomics, molecular biology, and forensic sciences applications. Dead Air Boxes with UV only are also available.
Brochure
Specification sheets