Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a range of high-quality TopAir clean benches, designed to supply a clean controlled work environment meeting Class 100/ISO5 cleanliness standards, resembling a clean room, with the additional advantages of portability and small dimensions. TopAir clean benches draw in air from the room or hall space, transfer the air through a HEPA filter using a fan, and subsequently clean the bench area with filtered air.
Clean benches provide a high-quality alternative to a clean room at a much lower cost and without any additional utility requirements.
Vertical PRO | Horizontal PRO | |
![]() | ![]() | |
Description | Air is channelled downwards through a filter installed at the top of the bench. Vertical air stream producing clean air at ISO-5/ CLASS100 or ISO4/Class10 standards. | Air flows through a filter installed at the back of the bench, toward the user. Horizontal air stream producing clean air in compliance with ISO5/ CLASS100 or ISO4/Class10 standards (depending on the filter installed). |
Advanced operation system |
| |
Red light alarm | Whenever there is an air velocity failure, a red light inside the unit switches on and the system light automatically switches off. This enables convenient detection of failures from a distance, even by hearing-impaired people. | |
Construction material |
| |
Sizes 4 standard sizes: |
|
|
Filter | HEPA Filter Efficiency of 99.9995% at 0.3 Microns H14 (Optional ULPA filter) | |
Optional accessories |
| |
Air velocity |
| |
Lighting | 800 LUX, LED lighting | |
Power supply | 230 V, 50/60 Hz, Single phase 230 V, 50/60 Hz, Single phase | |
Cleanliness within workstation | Class-100 (FS 209E) ISO 4, ISO-14644-1 | |
Request information › | Request information › |
Brochure