Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
PCR-UV or PCR-UV + HEPA cabinets offer a quality filtering system which provides complete protection for your samples from contamination in genomics, proteomics, molecular biology, and forensic sciences applications.
Made of high-quality non-corrosive polypropylene, the cabinets feature a high level of chemical resistance and are easy to clean. Polypropylene increases the product’s tensile strength and improves its thermal characteristics.
PCR-UV | PCR-UV+HEPA | |
Control panel | User-friendly control panel with UV, lighting control and UV timer (30 min) | Improved user-friendly 5″ control panel with UV, lighting control and UV timer (30 min) |
Safety features | UV safety mechanism Smart safety mechanism prevents UV exposure | |
Construction material | Polypropylene structure with high chemical resistance Tempered glass frameless pivot window | |
Sizes | 3 standard sizes: (external W x D x H)
| 3 standard sizes: (external W x D x H)
|
Filter | None | H14, HEPA Filter |
Air velocity | None | High-efficiency EC fan Wind speed at 0.3 m/s, 60 FPM |
Lighting | 17 W Ozone free UV lightbulb, UV output at 1 M 254 nm 800 LUX, Eco-friendly LED lighting | |
Power supply | 230 V, 50/60 Hz, Single phase | |
Test standard | Complies with Test Standard USA Federal Standard 209E/CE/ISO 1- 144641 | Complies with Test Standard USA Federal Standard 209E/CE/ISO 1- 144641 ISO 6/CLASS 1000 cleanliness level according to ISO 14644-1 and USA Federal Standard 209E, CE Certification |
Optional accessories | Metal stand | |
Request information › | Request information › |
Brochure
Specification sheets