Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
You have a choice of manual or automated pumps for transfer of fluids or gases. Options include vacuum pressure pumps, peristaltic pumps, water sampling pumps and more.
Pump type | Vacuum pump | Liquid dispensing |
---|---|---|
Rotary vane pump | ||
Diaphragm pump | ||
Syringe pump | ||
Peristaltic pump | ||
Scroll pump | ||
Drum pump | ||
Handheld vacuum pump | ||
Water sampling pumps |
Positive displacement roller pumps used to pump and transfer viscous gas or liquids in a variety of laboratory applications.
Pressure pump that extract gas or air to create a partial or complete vacuum in the system. Used for rotary evaporation, concentration, distillation, filtration and more.
Pneumatic pumps for dispensing, transferring or siphoning liquids from large carboys or drums. Can be used to with a pumping station.
Designed to meet EPA and NPDES guidelines, these peristaltic pumps are specifically used for environmental water sampling in groundwater, surface water, stormwater or wastewater.
Manually operated handheld pressure and vacuum pumps can be used as fluid aspirators that aid in filtration siphoning, preventing fluid line bleeding, checking for leaks, and liquid transfer.
Pumps used to gradually administer specific amounts of fluids for calibration, dilution, dosing, infusion, and more. Single-use, continuous cycle, high-pressure, and programmable/touch screen models are available.
Design the exact pump system you need. Masterflex has a broad range of pumps and fluid system components for laboratory, analytical, and industrial process applications. And not just pumps, they also offer tubing, fittings, regulators and gauges.
Includes a variety of tubing options for vacuum and liquid dispensing applications.
Pump replacement parts and repair service kits including pump heads, seals assemblies, drives and controllers.