Masterflex pumps
With over 55 years of manufacturing peristaltic pumps, Masterflex sets the standard for reliability and unparalleled versatility. Their experience makes them the fluid handling experts with the technical resources to help you design the exact pump system you need. Masterflex has a broad range of pumps and fluid system components for laboratory, analytical, and industrial process applications. And not just pumps, they also offer tubing, fittings, regulators and gauges.
Contact specialist
Benefits of choosing Masterflex
Masterflex tubing pump systems are perfect in the laboratory, manufacturing plant or field for the end-user. The convenient modular design of the systems (pump head + tubing + drive) allows you to use one pump drive for many applications, making Masterflex extremely versatile and cost-effective. The easy-loading pump head designs, coupled with a variety of specialty and general-use tubing formulations, make Masterflex the best choice for all your fluid handling needs.
  • Quick installation
  • Low maintenance
  • Easy tubing change over
  • Trusted for over 55 years
  • Extensive chemical compatibility
  • From research to process, handles almost any fluid
  • Multichannel capabilities
  • Configurable systems
  • Many tubing options
  • Easy scale-up
  • Flow ranges – 0.0001 mL/min to 42 L/min
  • Rigorous design standards
How to select your Masterflex pump system
Define your application requirements:
  • What flow rate do I need?
  • What chemical am I pumping?
  • Do conditions like fluid viscosity or back pressure play an essential role in my application?
Select a pump series based on your flow rate:
  • Masterflex Ismatec 0.002 to 37 mL/min
  • Masterflex L/S 0.0006 to 3400 mL/min
  • Masterflex I/P 0.012 to 13 LPM
  • Masterflex B/T 0.3 to 45 LPM
Select the components to fit your application:
  • I/P and L/S pumps systems are divided into three categories: pump head, tubing, and drive.
  • Masterflex Ismatec and B/T pumps systems are divided into two categories: pump head and drive.
  • To complete your tubing pump system, choose a pump head, tubing, and drive from within the same series.
What to consider
Masterflex Easy Load pump heads
Choosing a pump head
  • Flow rate: Different pump heads have different flow rate capabilities
  • Frequency of tubing changes: Some pump heads enable very rapid tubing changes
  • Desired number of channels: Pump heads are stackable in most cases
  • Chemical compatibility: The materials used for the pump head is important in order to combat any chemical spills
  • Accuracy: Standard pump heads deliver the most accurate and repeatable flow rates
Masterflex pump tubing
Choosing tubing
  • Size: Inner and outer diameter of the tubing determine flow range, maximum pressure, and ability to handle viscous fluids well.
  • Temperature: Higher fluid or environmental temperatures will decrease tubing life. Sometimes a layered tube will alleviate these concerns.
  • Application: Special considerations for your application of industry such as adherence to federal or other regulations such as FDA 21 cfr 177.2600 and USP Class VI, soft or hard particulates, or high viscosities are some examples.
  • Media: Choose the formulation that will be least affected by your fluid's chemical nature
  • Pressure: Higher pressures can result in decreased pump tubing life and increased possibility of rupture. This can be mitigated with thicker tube walls in conjunction with a pump head architecture that supports those pressure ranges. Additionally, tube or barb locks to fit over tubing connectors will help reduce the risk of rupture at those points
Masterflex digital drive
Choosing a drive
  • Flow rate: The rpm of the drive directly correlates to flow rate
  • Fixed- or variable-speed: Variable-speed drives enable flow rate adjustment
  • Accuracy of flow rate or dispense volume: Many drives have digital speed control circuitry for highly accurate and repeatable flow rates
  • Remote control via analog signal or cloud: Some drives can connect to your process controller or can be operated by a footswitch while others can be safely monitored or controlled by phone or webpage using the cloud
  • Environmental protection against hazards: Many drives are sealed against dust and water exposure
Masterflex peristaltic tubing pumps
Engineered for long life, Masterflex peristaltic pump systems provide superior accuracy and repeatability and ensure optimal performance when used with the proper tubing. These pumps offer the latest technology including MasterflexLive cloud-enabled connectivity, easy control and tubing changes, and the low-maintenance and cost-effectiveness of peristaltic pumps.
To provide you with an easier way to select the fine pumps we offer, Ismatec pumps are now a part of the Masterflex family of products. The Ismatec Series pumps fit into the Masterflex brand offering as the microflow, high accuracy peristaltic pumps. Choose from a wide range of flows with the Masterflex Ismatec Series, Masterflex L/S Series, Masterflex I/P Series and Masterflex B/T Series peristaltic pumps. You can also find many tubing formulations compatible with the pumps we offer. Keep in mind that Ismatec pumps accept Ismatec pump tubing and Masterflex pumps accept Masterflex pump tubing for clean fluid paths. With thousands of pump options and many tubing formulations, you can find what you need from the Masterflex brand.
Selecting a Masterflex pump is as easy as ABC
Application requirementsBase a pump series on your flow rateComplete your system
  • What is my required flow rate?
  • What chemical am I pumping?
  • Do conditions such as viscosity or back pressure play an important role in my decision?
  • Use the table below to find the pump series that best fits your flow rate needs
  • L/S and I/P pump series are divided into three components — pump head, tubing, and drive
  • Select the three components from the same series or choose a complete preconfigured system
Pump seriesFeatures
Flow
range
Fixed speedVariable speedRemote controlMultichannel capabilities
C/L®Compact, low-flow pump that easily fits on benchtop or mounts within a panel0.002 to 43 mL/min    
L/S®High-accuracy drives for precise flow control and dispensing0.0006 to 3400 mL/min    
I/P®Powerful systems that provide better protection from possible hazards such as dust and water0.012 to 19 L/min    
B/T®Highest flow rates — ideal for large batch fluid transfer with excellent hazard protection0.3 to 42 L/min    
Pumps and components
Masterflex pump systems
Complete Pump Systems
Complete pump systems include all the pump components you need to get pumping in no time at all. Our pump system includes a pump drive, pump head and pump tubing.
Peristaltic pump
Peristaltic Pumps
Masterflex peristaltic pump systems provide flexibility based on your needs and requirements. Masterflex pumps can be configured as your needs change — just switch out the pump head and the tubing to match your new requirements.
Masterflex Easy-Load pump head
Pump Heads
Standard pump heads offer the most accurate, repeatable flow rates.
Masterflex clear tubing
Peristaltic Pump Tubing
Precision extruded and pump tested formulations engineered to provide the highest quality for tubing life, pressure, and suction lift.
Masterflex vacuum pumps
Vacuum pumps, pressure pumps, cover a wide range of operating pressures. Laboratory/filtration vacuum pumps are for use in the lab or anywhere a low vacuum is needed; typically offer both vacuum and pressure capabilities. Use rough vacuum pumps (or roughing pumps) for laboratory and industrial applications requiring a level of vacuum less than 10–3 Torr. Use high vacuum pressure pumps when you need vacuums higher than 10–3 Torr. Also, consider the free-air capacity needed (the higher the free air capacity, the faster it will evacuate the chamber) and whether you need a lubricated (oiled) or non-lubricated (dry or oilless) pump. Lubricated pumps provide higher capacities, higher vacuum levels, and lower noise, but can contaminate the system and require more maintenance. Select a nonlubricated pump for vacuum in a clean system and low maintenance.
Masterflex Air Cadet vacuum pump
Air Cadet Vacuum/Pressure Pumps
Get better results with minimal dead space
  • Minimizes dead space and air entrapment with unique cavity configuration
  • No maintenance needed—AC motors are brushless with sealed ball bearings
  • Worry-free pumping with reduced stress, wear, and heat buildup
Masterflex Air Cadet PRO vacuum pump
Air Cadet PRO
Exceptional performance
  • General purpose pumps are great for filtration or vacuum drying
  • At 45 dB(A) it's quieter than a conversation
  • Long service intervals reduce maintenance costs
Designed for general laboratory applications, these pumps will give you thousands of hours of worry-free operation. Max vacuum of 29.8 in Hg. Pumps with gas ballast will allow purging of any contaminants that may condense in the pump head. A standard Air CadetPRO will work for all your basic vacuum needs including filtration and degassing. For demanding applications requiring solvents or corrosive chemicals use the chemical duty Air Cadet PRO.
Masterflex Air Admiral vacuum pump
Air Admiral Vacuum Pump and Station
Compact size and integral handle make pump easily transportable
  • Economical Air Admiral pump serves as pressure or vacuum source with an excellent price to performance ratio
  • Low-friction piston runs cool and quiet for a long service life with a consistent performance
This portable vacuum pump is designed for versatility with a tough, lightweight plastic housing and integral carrying handle. The long-wearing motor delivers performance characteristics suitable for a wide range of applications. Pump features a convenient cord storage compartment in its base.
Masterflex pump tubing
BioPharm Plus Tubing - Pharmaceuticals & Drug Discovery
Longer life (up to five times) and lower spallation compared to other platinum silicone tubing. Ultra-smooth inner surface. Biocompatible for sensitive applications. Autoclavable. Meets FDA, USP Class VI, and EP standards, and exceeds 3A sanitary standards. Max temp: 450°F (232°C)
Applications: filtration; fermentation; dispensing solutions and media.
  • Excellent pumping accuracy for high repeatability and consistent performance over time
  • Smooth inner bore results in less particle entrapment
  • Low spallation maintains purity of fluid
  • No leachable additives or plasticizers to impart taste or contamination
  • Sterilize to maintain system sterility
  • Tolerates high-pressure applications
C-FLEX® Tubing - Biopharmaceuticals & Nutraceuticals
Excellent biocompatibility; contains no toxic materials. Heat sealable and weldable. Autoclavable. Meets FDA and USP Class VI standards. Max temp: 275° F (135°C).
Applications: biopharmaceutical processing and storage; botanical production
  • Combines the biocompatibility of silicone with chemical resistance of Tygon®, but with longer pumping life
  • Nonpyrogenic, noncytotoxic, and nonhemolytic properties maintain purity and sterility of fluids
  • Low protein binding ensures minimal degradation or loss of sample
  • Chemically resistant to acids and bases for longer tubing life
PharmaPure® Tubing – Pharmaceutical, Life Science, Electronics & Semiconductor
Nontoxic and nonhemolytic, excellent biocompatibility. Longest pumping life of any high-performance formulation; low extractability. Autoclavable. Meets FDA, EP, and USP Class VI standards. Max temp: 275°F (135°C).
Applications: sterile filling; filtration; transfer of cell media.
  • Ultra-low spallation maintains fluid purity and extends the life of filters
  • Low permeability provides barrier to chemicals that damage tubing
  • Tolerates continuous-pressure applications up to 40 psi (2.7 bar)
  • Long tubing life means time and cost savings
Chem-Durance™ Tubing – Chemical, Printing & Automotive
Best combination of pumping life and chemical resistance. Excellent durability. Plasticizer-free inner liner provides low spallation. Autoclavable. Meets FDA standards. Max temp: 165°F (74°C).
Applications: transfer of sensitive fluids; ink and solvent production.
  • Chemical-resistant inner liner allows use with harsh chemicals
  • Low spallation maintains purity of fluid
  • Flexible outer jacket dissipates heat generated from rollers for longer tubing life
  • Hydrophobic, smooth-bore inner liner means negligible absorption of fluids or leachables
Pharmaceutical and Biotech
Pharmaceutical & biotech
Masterflex pumps are designed to be non-contaminating and non-invasive, making them the pumps of choice in the pharmaceutical industry. Easy-Load® pump heads and a wide range of drives fit many applications, from pumping nutrients or pH adjusters for fermentation to dispensing cosmetics into small containers. In addition, tubing is available that is compliant with USP Class VI, FDA, EP, and can be easily sterilised.
Research and Development
Research & development
Masterflex tubing pump systems are ideal for research and development. The pumps are very easy to load and extremely versatile for use in the laboratory or process scale-up areas. Repeatability is excellent for low-volume dispensing and metering applications, and, under most conditions, the valveless design eliminates clogging and siphoning of fluid. In addition, these modular systems allow you to use one pump drive for many applications.
Industrial and Manufacturing
Industrial & manufacturing
For everyday transferring and metering applications in the laboratory or plant, Masterflex pumps are the answer. They offer predictable service in continuous-duty areas while solving many tough pumping applications. Use these pumps to handle wastewater, suspended solids, harsh chemicals and more; up to 42 LPM (11.1 GPM). The pumps are easy to use and maintain, and the drives are designed to handle the rugged environment of any plant.
Food and Beverage
Food & beverage
Masterflex can handle a wide range of fluids, from hot water and vitamin solutions to thick molasses. The pump heads are designed for easy tubing exchange, and a wide selection of the drives have ”wash-down” capabilities. Most importantly, Masterflex tubing is available for food applications that meets USP Class VI, FDA, NSF, EP, and 3A requirements.
environmental_230x195px
Environmental & special
Portability and convenience are built into every Masterflex environmental sampling pump. Choices of DC or AC systems facilitate simple, accurate sample collection. In addition, the suction lift and non-invasive/non-contaminating designs of the pump heads and tubing enable sample collection under EPA and other standard protocols.
How to select your Masterflex pump system
Define your application requirements:
  • What flow rate do I need?
  • What chemical am I pumping?
  • Do conditions like fluid viscosity or back pressure play an essential role in my application?
Select a pump series based on your flow rate:
  • Masterflex Ismatec 0.002 to 37 mL/min
  • Masterflex L/S 0.0006 to 3400 mL/min
  • Masterflex I/P 0.012 to 13 LPM
  • Masterflex B/T 0.3 to 45 LPM
Select the components to fit your application:
  • I/P and L/S pumps systems are divided into three categories: pump head, tubing, and drive.
  • Masterflex Ismatec and B/T pumps systems are divided into two categories: pump head and drive.
  • To complete your tubing pump system, choose a pump head, tubing, and drive from within the same series.
What to consider
Masterflex Easy Load pump heads
Choosing a pump head
  • Flow rate: Different pump heads have different flow rate capabilities
  • Frequency of tubing changes: Some pump heads enable very rapid tubing changes
  • Desired number of channels: Pump heads are stackable in most cases
  • Chemical compatibility: The materials used for the pump head is important in order to combat any chemical spills
  • Accuracy: Standard pump heads deliver the most accurate and repeatable flow rates
Masterflex pump tubing
Choosing tubing
  • Size: Inner and outer diameter of the tubing determine flow range, maximum pressure, and ability to handle viscous fluids well.
  • Temperature: Higher fluid or environmental temperatures will decrease tubing life. Sometimes a layered tube will alleviate these concerns.
  • Application: Special considerations for your application of industry such as adherence to federal or other regulations such as FDA 21 cfr 177.2600 and USP Class VI, soft or hard particulates, or high viscosities are some examples.
  • Media: Choose the formulation that will be least affected by your fluid's chemical nature
  • Pressure: Higher pressures can result in decreased pump tubing life and increased possibility of rupture. This can be mitigated with thicker tube walls in conjunction with a pump head architecture that supports those pressure ranges. Additionally, tube or barb locks to fit over tubing connectors will help reduce the risk of rupture at those points
Masterflex digital drive
Choosing a drive
  • Flow rate: The rpm of the drive directly correlates to flow rate
  • Fixed- or variable-speed: Variable-speed drives enable flow rate adjustment
  • Accuracy of flow rate or dispense volume: Many drives have digital speed control circuitry for highly accurate and repeatable flow rates
  • Remote control via analog signal or cloud: Some drives can connect to your process controller or can be operated by a footswitch while others can be safely monitored or controlled by phone or webpage using the cloud
  • Environmental protection against hazards: Many drives are sealed against dust and water exposure
Masterflex peristaltic tubing pumps
Engineered for long life, Masterflex peristaltic pump systems provide superior accuracy and repeatability and ensure optimal performance when used with the proper tubing. These pumps offer the latest technology including MasterflexLive cloud-enabled connectivity, easy control and tubing changes, and the low-maintenance and cost-effectiveness of peristaltic pumps.
To provide you with an easier way to select the fine pumps we offer, Ismatec pumps are now a part of the Masterflex family of products. The Ismatec Series pumps fit into the Masterflex brand offering as the microflow, high accuracy peristaltic pumps. Choose from a wide range of flows with the Masterflex Ismatec Series, Masterflex L/S Series, Masterflex I/P Series and Masterflex B/T Series peristaltic pumps. You can also find many tubing formulations compatible with the pumps we offer. Keep in mind that Ismatec pumps accept Ismatec pump tubing and Masterflex pumps accept Masterflex pump tubing for clean fluid paths. With thousands of pump options and many tubing formulations, you can find what you need from the Masterflex brand.
Selecting a Masterflex pump is as easy as ABC
Application requirementsBase a pump series on your flow rateComplete your system
  • What is my required flow rate?
  • What chemical am I pumping?
  • Do conditions such as viscosity or back pressure play an important role in my decision?
  • Use the table below to find the pump series that best fits your flow rate needs
  • L/S and I/P pump series are divided into three components — pump head, tubing, and drive
  • Select the three components from the same series or choose a complete preconfigured system
Pump seriesFeatures
Flow
range
Fixed speedVariable speedRemote controlMultichannel capabilities
C/L®Compact, low-flow pump that easily fits on benchtop or mounts within a panel0.002 to 43 mL/min    
L/S®High-accuracy drives for precise flow control and dispensing0.0006 to 3400 mL/min    
I/P®Powerful systems that provide better protection from possible hazards such as dust and water0.012 to 19 L/min    
B/T®Highest flow rates — ideal for large batch fluid transfer with excellent hazard protection0.3 to 42 L/min    
Pumps and components
Masterflex pump systems
Complete Pump Systems
Complete pump systems include all the pump components you need to get pumping in no time at all. Our pump system includes a pump drive, pump head and pump tubing.
Peristaltic pump
Peristaltic Pumps
Masterflex peristaltic pump systems provide flexibility based on your needs and requirements. Masterflex pumps can be configured as your needs change — just switch out the pump head and the tubing to match your new requirements.
Masterflex Easy-Load pump head
Pump Heads
Standard pump heads offer the most accurate, repeatable flow rates.
Masterflex clear tubing
Peristaltic Pump Tubing
Precision extruded and pump tested formulations engineered to provide the highest quality for tubing life, pressure, and suction lift.
Masterflex vacuum pumps
Vacuum pumps, pressure pumps, cover a wide range of operating pressures. Laboratory/filtration vacuum pumps are for use in the lab or anywhere a low vacuum is needed; typically offer both vacuum and pressure capabilities. Use rough vacuum pumps (or roughing pumps) for laboratory and industrial applications requiring a level of vacuum less than 10–3 Torr. Use high vacuum pressure pumps when you need vacuums higher than 10–3 Torr. Also, consider the free-air capacity needed (the higher the free air capacity, the faster it will evacuate the chamber) and whether you need a lubricated (oiled) or non-lubricated (dry or oilless) pump. Lubricated pumps provide higher capacities, higher vacuum levels, and lower noise, but can contaminate the system and require more maintenance. Select a nonlubricated pump for vacuum in a clean system and low maintenance.
Masterflex Air Cadet vacuum pump
Air Cadet Vacuum/Pressure Pumps
Get better results with minimal dead space
  • Minimizes dead space and air entrapment with unique cavity configuration
  • No maintenance needed—AC motors are brushless with sealed ball bearings
  • Worry-free pumping with reduced stress, wear, and heat buildup
Masterflex Air Cadet PRO vacuum pump
Air Cadet PRO
Exceptional performance
  • General purpose pumps are great for filtration or vacuum drying
  • At 45 dB(A) it's quieter than a conversation
  • Long service intervals reduce maintenance costs
Designed for general laboratory applications, these pumps will give you thousands of hours of worry-free operation. Max vacuum of 29.8 in Hg. Pumps with gas ballast will allow purging of any contaminants that may condense in the pump head. A standard Air CadetPRO will work for all your basic vacuum needs including filtration and degassing. For demanding applications requiring solvents or corrosive chemicals use the chemical duty Air Cadet PRO.
Masterflex Air Admiral vacuum pump
Air Admiral Vacuum Pump and Station
Compact size and integral handle make pump easily transportable
  • Economical Air Admiral pump serves as pressure or vacuum source with an excellent price to performance ratio
  • Low-friction piston runs cool and quiet for a long service life with a consistent performance
This portable vacuum pump is designed for versatility with a tough, lightweight plastic housing and integral carrying handle. The long-wearing motor delivers performance characteristics suitable for a wide range of applications. Pump features a convenient cord storage compartment in its base.
Masterflex pump tubing
BioPharm Plus Tubing - Pharmaceuticals & Drug Discovery
Longer life (up to five times) and lower spallation compared to other platinum silicone tubing. Ultra-smooth inner surface. Biocompatible for sensitive applications. Autoclavable. Meets FDA, USP Class VI, and EP standards, and exceeds 3A sanitary standards. Max temp: 450°F (232°C)
Applications: filtration; fermentation; dispensing solutions and media.
  • Excellent pumping accuracy for high repeatability and consistent performance over time
  • Smooth inner bore results in less particle entrapment
  • Low spallation maintains purity of fluid
  • No leachable additives or plasticizers to impart taste or contamination
  • Sterilize to maintain system sterility
  • Tolerates high-pressure applications
C-FLEX® Tubing - Biopharmaceuticals & Nutraceuticals
Excellent biocompatibility; contains no toxic materials. Heat sealable and weldable. Autoclavable. Meets FDA and USP Class VI standards. Max temp: 275° F (135°C).
Applications: biopharmaceutical processing and storage; botanical production
  • Combines the biocompatibility of silicone with chemical resistance of Tygon®, but with longer pumping life
  • Nonpyrogenic, noncytotoxic, and nonhemolytic properties maintain purity and sterility of fluids
  • Low protein binding ensures minimal degradation or loss of sample
  • Chemically resistant to acids and bases for longer tubing life
PharmaPure® Tubing – Pharmaceutical, Life Science, Electronics & Semiconductor
Nontoxic and nonhemolytic, excellent biocompatibility. Longest pumping life of any high-performance formulation; low extractability. Autoclavable. Meets FDA, EP, and USP Class VI standards. Max temp: 275°F (135°C).
Applications: sterile filling; filtration; transfer of cell media.
  • Ultra-low spallation maintains fluid purity and extends the life of filters
  • Low permeability provides barrier to chemicals that damage tubing
  • Tolerates continuous-pressure applications up to 40 psi (2.7 bar)
  • Long tubing life means time and cost savings
Chem-Durance™ Tubing – Chemical, Printing & Automotive
Best combination of pumping life and chemical resistance. Excellent durability. Plasticizer-free inner liner provides low spallation. Autoclavable. Meets FDA standards. Max temp: 165°F (74°C).
Applications: transfer of sensitive fluids; ink and solvent production.
  • Chemical-resistant inner liner allows use with harsh chemicals
  • Low spallation maintains purity of fluid
  • Flexible outer jacket dissipates heat generated from rollers for longer tubing life
  • Hydrophobic, smooth-bore inner liner means negligible absorption of fluids or leachables
Pharmaceutical and Biotech
Pharmaceutical & biotech
Masterflex pumps are designed to be non-contaminating and non-invasive, making them the pumps of choice in the pharmaceutical industry. Easy-Load® pump heads and a wide range of drives fit many applications, from pumping nutrients or pH adjusters for fermentation to dispensing cosmetics into small containers. In addition, tubing is available that is compliant with USP Class VI, FDA, EP, and can be easily sterilised.
Research and Development
Research & development
Masterflex tubing pump systems are ideal for research and development. The pumps are very easy to load and extremely versatile for use in the laboratory or process scale-up areas. Repeatability is excellent for low-volume dispensing and metering applications, and, under most conditions, the valveless design eliminates clogging and siphoning of fluid. In addition, these modular systems allow you to use one pump drive for many applications.
Industrial and Manufacturing
Industrial & manufacturing
For everyday transferring and metering applications in the laboratory or plant, Masterflex pumps are the answer. They offer predictable service in continuous-duty areas while solving many tough pumping applications. Use these pumps to handle wastewater, suspended solids, harsh chemicals and more; up to 42 LPM (11.1 GPM). The pumps are easy to use and maintain, and the drives are designed to handle the rugged environment of any plant.
Food and Beverage
Food & beverage
Masterflex can handle a wide range of fluids, from hot water and vitamin solutions to thick molasses. The pump heads are designed for easy tubing exchange, and a wide selection of the drives have ”wash-down” capabilities. Most importantly, Masterflex tubing is available for food applications that meets USP Class VI, FDA, NSF, EP, and 3A requirements.
environmental_230x195px
Environmental & special
Portability and convenience are built into every Masterflex environmental sampling pump. Choices of DC or AC systems facilitate simple, accurate sample collection. In addition, the suction lift and non-invasive/non-contaminating designs of the pump heads and tubing enable sample collection under EPA and other standard protocols.
Style Sheet for Global Design System