Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
High capacity modular systems can accommodate a large number of samples in a wide variety of formats. These systems offer the flexibility of working with 26 different rotors, including rotor blocks, to fit a variety of sample formats such as microcentrifuge tubes, vials, microwell plates and flasks for your application needs.
SPD210 | SPD300DDA | |
---|---|---|
Cover lid construction | Glass | Glass heated cover |
Chamber heater assembly | Yes | Yes (4 radiant lamps) |
Heated lid (halogen lamp) | NA | Yes |
Vacuum ramp | No | Yes |
Halogen lamp in chamber | NA | Yes (4) |
Chamber temp operating range | Ambient -65°C | Ambient to 80°C |
Vacuum connection port | 0.5" | 2" |
User interface | ||
Total # of programs | 3 | 12 |
Presets | 3 | 4 |
Solvent types | EtOH, H20, buffers, Ammonium hydroxide (oligos), DNA in acetonitrile, acetonitrile, methanol, ethanol + .1% TFA | EtOH, H20, buffers, Ammonium hydroxide (oligos), DNA in acetonitrile, acetonitrile, methanol, ethanol + .1% TFA, Chloroform, ethyl acetate, acetone, methylene chloride, hexane, toluene, nitrobenzene, pyridine, TFA, HCL, acetic acid, formic acid, sodium hydroxide |
User defined programs | 3 | 3 |
Vacuum control | No | Yes |
Applications | Proteomics, chromatography, life sciences | Genomics, drug discovery, proteomics, chromatography, life sciences |
Available kits | ||
Refrigerated vapor trap kits | SPD210P1-115: 115V/60Hz
| SPD300P1-115: 115V/60Hz
|
Universal vacuum system kits | SPD210P2-115: 115V/60Hz
| SPD300P2-115: 115V/60Hz
|