For ease of use and speed of evaporation
SPD130DLX has radiant cover to improve drying times by applying radiant heat from the top of the samples.
Built in vacuum ramp and vacuum gauge in SPD130DLX and SPD140DDA allows adjustable vacuum level which prevents bumping and optimize drying time. View all small & medium capacity SpeedVac rotors
SpeedVac Selection Guide
SPD120
SPD130DLX
SPD140DDA
Cover lid constructionGlassRadiant glassGlass heated cover
Chamber heater assembly  YesYesYes (2 radiant lamps)
Heated lid (halogen lamp)NAYesNA
Vacuum rampNANAYes
Halogen lamp in chamberNANAYes (2)
User interface
Total # of programs3612
Presets334
Solvent typesWater, low boiling solvents (acetonitrile / methanol), HPLC / water, low boiling acidWater, low boiling solvents (acetonitrile / methanol), HPLC / water, low boiling acidWater, high and low boiling solvents (acetic acid, isopropanol, toluene, DMSO, TFA), HPLC / water
User defined programs338
Vacuum controlNoYesYes
ApplicationsDNA, RNA, proteinsOrganic chemistryOrganic chemistry, acid resistant
Available kits
Refrigerated vapor trap kits
SPD120P2-115: 115V/60Hz
SPD120P2-230: 230V/50Hz
  • SpeedVac concentrator
  • -105ºC, ultra-low temperature refrigerated vapor trap (RVT5105)
  • VPL120 high vacuum pump
  • 40 x 1.5mL rotor (RH40-11)
  • Universal tubing package (TFK100)
  • Digital vacuum gauge
  • Chemical trap kit (SCT120)
  • Disposable cartridge for trapping acids and water vapor (DC120A)
  • Glass condensation flask (GCF400)
  • Vacuum pump lubricating oil
  • CryoCool heat transfer fluid (1L)
SPD130P1-115: 115V/60Hz
SPD130P1-230: 230V/50Hz
  • SpeedVac concentrator
  • -105ºC, ultra-low temperature refrigerated vapor trap (RVT5105)
  • OFP400 Oil-Free ETFE/PFA 4-Headed Pump
  • 64 × 1.5mL Rotor (RH64-11)
  • Tubing package
  • Digital vacuum gauge
  • Chemical trap kit (SCT120)
  • Disposable cartridge for trapping acids and water vapor (DC120A)
  • Glass condensation flask (GCF400)
  • CryoCool heat transfer fluid (1L)
SPD140P1-115: 115V/60Hz
SPD140P1-230: 230V/50Hz
  • Acid-Resistant SpeedVac concentrator
  • -104°C Ultra-Low refrigerated vapor trap (RVT5105)
  • Oil-Free ETFE/PFA 4-Headed Pump
  • 40 x 1.5-2.0mL glass tube rotor (RH40-11)
  • Glass condensation flask (GCF400)
  • Tubing and fitting kit
  • CryoCool heat transfer fluid (1L)
Universal vacuum system kits
SPD120P1-115: 115V/60Hz
SPD120P1-230: 230V/50Hz
  • SpeedVac concentrator
  • Universal vacuum system (UVS450) including an oil-free vacuum pump and a -50°C refrigerated vapor trap
  • Glass condensation flask (GCF400)
  • 40 x 1.5mL rotor (RH40-11)
  • Tubing package
  • CryoCool heat transfer fluid (1L)
SPD140P2-115: 115V/60Hz
SPD140P2-230: 230V/50Hz
  • Acid-Resistant SpeedVac concentrator
  • -50°C Universal Vacuum Source
  • 24 x 1 dram vials (15 x 45mm) 4mL - dram vial Rotor (RH24-15)
  • Chemical trap (SCT120) and disposable cartridge (DC120A)
  • Tubing and fitting kit
  • Glass condensation flask (GCF400)
  • Vacuum pump lubricating oil
  • CryoCool heat transfer fluid (1L)