Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
SPD130DLX has radiant cover to improve drying times by applying radiant heat from the top of the samples.
Built in vacuum ramp and vacuum gauge in SPD130DLX and SPD140DDA allows adjustable vacuum level which prevents bumping and optimize drying time. View all small & medium capacity SpeedVac rotors
SPD120 | SPD130DLX | SPD140DDA | |
---|---|---|---|
Cover lid construction | Glass | Radiant glass | Glass heated cover |
Chamber heater assembly | Yes | Yes | Yes (2 radiant lamps) |
Heated lid (halogen lamp) | NA | Yes | NA |
Vacuum ramp | NA | NA | Yes |
Halogen lamp in chamber | NA | NA | Yes (2) |
User interface | |||
Total # of programs | 3 | 6 | 12 |
Presets | 3 | 3 | 4 |
Solvent types | Water, low boiling solvents (acetonitrile / methanol), HPLC / water, low boiling acid | Water, low boiling solvents (acetonitrile / methanol), HPLC / water, low boiling acid | Water, high and low boiling solvents (acetic acid, isopropanol, toluene, DMSO, TFA), HPLC / water |
User defined programs | 3 | 3 | 8 |
Vacuum control | No | Yes | Yes |
Applications | DNA, RNA, proteins | Organic chemistry | Organic chemistry, acid resistant |
Available kits | |||
Refrigerated vapor trap kits | SPD120P2-115: 115V/60Hz
| SPD130P1-115: 115V/60Hz
| SPD140P1-115: 115V/60Hz
|
Universal vacuum system kits | SPD120P1-115: 115V/60Hz
| SPD140P2-115: 115V/60Hz
|