Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Enhance air disinfection in your work environment with Grant Instruments Air Flow Cleaners. These easy-to-use UV cleaner-recirculators are ideal for use in hospitals (outpatient departments, operating rooms, emergency rooms, delivery rooms etc.), research laboratories, veterinary clinics, schools, and offices. They can be either fixed on walls (standard) or mounted on a movable tripod (optional).
GRAUVR-M | GRAUVR-Mi | GRAUV360 (AP 360) | |
---|---|---|---|
UV radiation source | 1 lamp (25 W Bactericidal, TUV25WG13 UV-C) | 2 lamp (25 W Bactericidal, TUV25WG13 UV-C) | 2 lamps (55 W Bactericidal, TU PL-L 55W UV-C) |
UV radiation level | 18 mW/cm2/sec | 36 mW/cm2/sec | 156 mW/cm2/sec |
Air-flow productivity | 14 m3/hour | 14 m3/hour | 360 m3/hour |
Full user protection from direct UV light | ✓ | ✓ | ✓ |
Display | — | LCD | LCD |
Timer | — | 1 min to 24 hours/non-stop | 1 min to 24 hours/non-stop |
UV lamp operation indicator | ✓ | ✓ | ✓ |
UV lamp lifetime counter | — | ✓ | ✓ |
Automatic switch ON/OFF | — | ✓ | ✓ |
Lamp fault detection | — | ✓ | ✓ |
Nominal operating voltage | 230 V, 50 Hz | 230 V, 50 Hz | 230 V, 50 Hz |
Power consumption (230 V) | 125 VA (540 mA) | 125 VA (540 mA) | Low |
Overall dimensions (h x d x w) | 660 mm x 135 mm x 110 mm | 660 mm x 135 mm x 110 mm | 1050 mm x 350 mm x 900 mm |
Weight | 3.4 kg | 3.4 kg | 13.4 kg |