Grant Instruments Air Flow Cleaner
Enhance air disinfection in your work environment with Grant Instruments Air Flow Cleaners. These easy-to-use UV cleaner-recirculators are ideal for use in hospitals (outpatient departments, operating rooms, emergency rooms, delivery rooms etc.), research laboratories, veterinary clinics, schools, and offices. They can be either fixed on walls (standard) or mounted on a movable tripod (optional).
Request information
Compare Grant Instruments air flow cleaners
 GRAUVR-MGRAUVR-MiGRAUV360 (AP 360)
UV radiation source 1 lamp (25 W Bactericidal, TUV25WG13 UV-C)2 lamp (25 W Bactericidal, TUV25WG13 UV-C)2 lamps (55 W Bactericidal, TU PL-L 55W UV-C)
UV radiation level18 mW/cm2/sec36 mW/cm2/sec156 mW/cm2/sec
Air-flow productivity14 m3/hour14 m3/hour360 m3/hour
Full user protection from direct UV light
DisplayLCDLCD
Timer1 min to 24 hours/non-stop1 min to 24 hours/non-stop
UV lamp operation indicator
UV lamp lifetime counter
Automatic switch ON/OFF
Lamp fault detection
Nominal operating voltage230 V, 50 Hz230 V, 50 Hz230 V, 50 Hz
Power consumption (230 V)125 VA (540 mA)125 VA (540 mA) Low
Overall dimensions (h x d x w)660 mm x 135 mm x
110 mm
660 mm x 135 mm x
110 mm
1050 mm x 350 mm x
900 mm
Weight3.4 kg3.4 kg13.4 kg